Skip to Content
Merck
All Photos(1)

Key Documents

5.43835

Supelco

Ammonium hydrogen carbonate

for HPLC LiChropur

Synonym(s):

Ammonium hydrogen carbonate, Ammonium bicarbonate, Ammonium carbonate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$137.00
250 G
$303.00

$137.00


Estimated to ship on07 April 2025


Request a Bulk Order

Select a Size

Change View
100 G
$137.00
250 G
$303.00

About This Item

Linear Formula:
(NH4)HCO3
CAS Number:
Molecular Weight:
79.06
MDL number:
UNSPSC Code:
12352301
EC Index Number:
213-911-5
NACRES:
NA.21

$137.00


Estimated to ship on07 April 2025


Request a Bulk Order

vapor pressure

67 hPa ( 20 °C)

Quality Level

form

solid

pH

8 (20 °C, 50 g/L in H2O)

mp

106 °C

solubility

220 g/L

storage temp.

2-30°C

SMILES string

[Zn+2].[Zn+2].N.N.[O-]C(=O)[O-].[O-]C(=O)[O-].OC(=O)O

InChI

1S/3CH2O3.2H3N.2Zn/c3*2-1(3)4;;;;/h3*(H2,2,3,4);2*1H3;;/q;;;;;2*+2/p-4

InChI key

QOPSSYZLVIENBR-UHFFFAOYSA-J

General description

Ammonium hydrogen carbonate is used in HPLCanalysis as a mobile phase component. It is a volatile buffer salt that canprovide pH control and buffering capacity in HPLC separations.[1]

Application


  • Aluminon: Its Limited Application as a Reagent for the Detection of Aluminum Species: This research explores the application limitations of Aurin tricarboxylic acid ammonium salt as a reagent in detecting aluminum species, providing critical insights into its specificity and interaction with metal ions (Clark RA, Krueger GL, 1985).


Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suppression effects of carbonate on the interaction between stainless steel and phosphate groups of phosphate compounds in high-performance liquid chromatography and electrospray ionization mass spectrometry
Asakawa Y, et al.
Journal of Chromatography A, 1198, 80-86 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service