Skip to Content
Merck
All Photos(1)

Key Documents

28638-U

Hamilton® 1002LTN syringe

1002LTN, volume 2.5 mL, needle size 22 ga (blunt tip), needle L 51 mm (2 in.)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$181.00

$181.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$181.00

About This Item

UNSPSC Code:
41122001
NACRES:
NB.03

$181.00


Check Cart for Availability

Request a Bulk Order

material

blunt tip stainless steel (point style 3)

needle size

22 ga (blunt tip)

description

gastight

feature

GASTIGHT® Syringe, Fixed needle

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 203083

needle L

51 mm (2 in.)

volume

2.5 mL

compatibility

for use with CTC HTS PAL Instruments (HPLC Sampling)

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. We need of the picture. Is it possible?

    1 answer
    1. Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service