Skip to Content
Merck
All Photos(1)

Key Documents

20890-U

Hamilton® HPLC syringes

1750RNR, volume 0.5 mL, needle size 22 ga (blunt tip), needle L 51 mm (2 in.)

Synonym(s):

Analytical HPLC Injection Syringes

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$146.00

$146.00


Available to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 EA
$146.00

About This Item

UNSPSC Code:
41122001
NACRES:
NB.03

$146.00


Available to ship on11 April 2025Details


Request a Bulk Order

material

blunt tip stainless steel needle (point style 3)
glass body

needle size

22 ga (blunt tip)

feature

blunt tip

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 81265

needle L

51 mm (2 in.)

volume

0.5 mL

compatibility

for use with Rheodyne
for use with SSI
for use with Valco VISF-2

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Hamilton is a registered trademark of Hamilton Co.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service