Skip to Content
Merck
All Photos(8)

Key Documents

EZFITBASE6

Millipore

EZ-Fit® Manifold

316L stainless steel, 6-place manifold base for filtration application

Synonym(s):

EZ-Fit® Manifold base, EZ-Fit® manifold base for EZ-Stream® vacuum pump

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$8,130.00

$8,130.00


Available to ship on22 April 2025Details


Request more information

Select a Size

Change View
1 EA
$8,130.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

$8,130.00


Available to ship on22 April 2025Details


Request more information

Quality Level

material

316L stainless steel

Sterilization Compatibility

autoclavable compatible

feature

6-place manifold base for filtration application

packaging

pkg of 1 unit

manufacturer/tradename

EZ-Fit®

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

for use with all EZ-Fit® Filtration heads (EZ-Fit®, Microfil®, Milliflex Oasis®)

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

EZ-FIT® vacuum manifold is a state-of-the-art system supporting the sample filtration. Millipore glass, plastic or stainless-steel filter holders can be fitted with standard No. 8 perforated stoppers over holder outlets. Different heads fit the disposable consumables, as well as the reusable funnels. The manifolds optimize the workflow, control the contamination risks, and avoid false-positive results during contamination tests. The design of the 2-way valve prevents accidental touching of the filtration head while opening or closing the valve. The liquid back-flow from the vacuum system is avoided by the check valve. The low height design offers better ergonomics for use in laminar flow hoods.

Application

EZ-FIT® vacuum manifolds are used as vacuum pump accessories to filter the test samples for general microbiological analysis, bioburden testing (pharmaceuticals, food and beverage) and water monitoring. It supports the filtration of six test samples.

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
EZ-STREAM is a registered trademark of Merck KGaA, Darmstadt, Germany
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service