Skip to Content
Merck
All Photos(1)

Key Documents

EZFITGASK3

Millipore

EZ-Fit® Manifold o-ring kit

for use with EZ-Fit® Filtration Manifold

Sign Into View Organizational & Contract Pricing

Select a Size

3 EA
$119.00

$119.00


Check Cart for Availability

Request more information

Select a Size

Change View
3 EA
$119.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

$119.00


Check Cart for Availability

Request more information

material

EPDM rubber

Quality Level

Sterilization Compatibility

autoclavable compatible

packaging

pkg of 3 pieces

manufacturer/tradename

EZ-Fit®

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

for use with EZ-Fit® Filtration Manifold

shipped in

ambient

Related Categories

General description

o-ring kit is an additional accessory for use with EZ-Fit® Manifold. EZ-Fit® vacuum manifold is a state-of-the-art system supporting the sample filtration. The manifolds optimize the workflow, control the contamination risks, and avoid false-positive results during contamination tests. With the EZ-Fit® Manifold, biofilm formation can be prevented because every single component can be removed for easy cleaning and autoclaving. The o-rings help in tight sealing of the funnel to the manifold thereby, aids in the leak-free operation and uniform microorganism recoveries.

Application

O- ring kit is an additional accessory for use with EZ-Fit® Manifold that helps in tight sealing of the funnel to the manifold thereby, aids in the leak-free operation and uniform microorganism recoveries from beer, bottled water, and pharmaceutical products.

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service