Skip to Content
Merck
All Photos(1)

Documents

5.06384

Sigma-Aldrich

BLM Helicase Inhibitor, ML216

Synonym(s):

BLM Helicase Inhibitor, ML216, 1-(4-Fluoro-3-(trifluoromethyl)phenyl)-3-(5-(pyridin-4-yl)-1,3,4-thiadiazol-2-yl)urea, ML-216, ML-216, 1-(4-Fluoro-3-(trifluoromethyl)phenyl)-3-(5-(pyridin-4-yl)-1,3,4-thiadiazol-2-yl)urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H9F4N5OS
CAS Number:
Molecular Weight:
383.32
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 10 mg/mL

storage temp.

2-8°C

General description

A cell-permeable pyridinyl-thiadiazolylurea compound that selectively blocks Bloom′s syndrome protein (BLM) helicase activity of both full-length as well as truncated BLM636-1298 forms (IC50 = 2.98 and 0.97 µM, respectively). Competes with and reversibly blocks DNA binding to BLM. However, the inhibition appears to be non-competitive with respect to ATP. Exhibits anti-proliferative activity in cells expressing BLM, but not in those lacking BLM. Induces sister chromatid exchanges (SCEs) and enhances the sensitivity of BLM containing PSNF5 cells to aphidicolin (>Cat. No. 178273) without affecting PSNG13 cells that are devoid of BLM. Displays favorable microsomal (T1/2 >1.2 h) and plasma stability (~100% at 5 h).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
BLM
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Rosenthal, A.S., et al. 2013. Bioorg. Med. Chem. Lett. 23, 5660.
Nguyen, G.H., et al. 2013. Chem. Biol.20, 55.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service