Skip to Content
Merck
All Photos(1)

Key Documents

5.00503

Sigma-Aldrich

CDC42 Inhibitor III, ZCL278

Synonym(s):

CDC42 Inhibitor III, ZCL278, CDC42 GTPase Inhibitor III, 4-(3-(2-(4-Bromo-2-chloro-phenoxy)-acetyl)-thioureido)-N-(4,6-dimethyl-pyrimidin-2-yl)-benzenesulfonamide, 2-(4-Bromo-2-chlorophenoxy)-N-((4-((4,6-dimethyl-2-pyrimidinyl)sulfamoyl)phenyl)carbamothioyl)acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$314.00

$314.00


Estimated to ship on30 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
25 MG
$314.00

About This Item

Empirical Formula (Hill Notation):
C21H19BrClN5O4S2
CAS Number:
Molecular Weight:
584.89
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$314.00


Estimated to ship on30 May 2025

New, lower price on this item!

Request a Bulk Order

Assay

≥94% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light beige

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(Nc3nc(cc(n3)C)C)c1ccc(cc1)NC(=S)NC(=O)COc2c(cc(cc2)Br)Cl

InChI

1S/C21H19BrClN5O4S2/c1-12-9-13(2)25-20(24-12)28-34(30,31)16-6-4-15(5-7-16)26-21(33)27-19(29)11-32-18-8-3-14(22)10-17(18)23/h3-10H,11H2,1-2H3,(H,24,25,28)(H2,26,27,29,33)

InChI key

XKZDWYDHEBCGCG-UHFFFAOYSA-N

General description

A cell-permeable, non-cytotoxic, aqueous soluble (181 µM in PBS, pH 7.4, containing 0.33% DMSO) pyrimidinylthiourea compound that targets Cdc42 GEF ITSN/intersectin and adjacent GTP/GDP binding site (Kd = 6.4 and 11.4 µM by fluorescence titration and SPR, respectively) and effectively inhibits EGF-stimulated Cdc42 activity (by 100% with 1 h pretreatment of 50 µM ZCL278) and Cdc42-dependent microspike/filopodia formation in serum-starved Swiss 3T3 fibroblast cultures without affecting RhoA-dependent cellular processes or Rac1-dependent lamellipodia formation. Short-term ZCL278 exposure in 5 d-cultured primary neonatal cortical neurons from 1 d postnatal mice is shown to dramatically abolish neurite branching (Ave branches per neurite = 21, 7.8, and 6.4, respectively, with 0, 5, and 10 min exposure to 50 µM ZCL278).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CDC42
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze 9-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Friesland, A., et al. 2013. Proc. Natl. Acad. Sci. USA110, 1261.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenjie Huang et al.
Cancer research, 80(13), 2804-2817 (2020-05-08)
Pancreatic ductal adenocarcinoma (PDAC) is an aggressive malignancy characterized by extensive local invasion and systemic spread. In this study, we employed a three-dimensional organoid model of human pancreatic cancer to characterize the molecular alterations critical for invasion. Time-lapse microscopy was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service