Skip to Content
Merck
All Photos(1)

Key Documents

681647

Sigma-Aldrich

WRN Helicase Inhibitor, NSC 19630

The WRN Helicase Inhibitor, NSC 19630, also referenced under CAS 72835-26-8, controls the biological activity of WRN Helicase. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

WRN Helicase Inhibitor, NSC 19630, 1-((1-Oxopropoxy)methyl)-maleimide, 1-((1-Oxopropoxy)methyl)-1H-pyrrole-2,5-dione, MIRA-1, p53 Activator VIII

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$211.00

$211.00


Estimated to ship on31 May 2025



Select a Size

Change View
5 MG
$211.00

About This Item

Empirical Formula (Hill Notation):
C8H9NO4
CAS Number:
Molecular Weight:
183.16
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$211.00


Estimated to ship on31 May 2025


Quality Level

Assay

≥97% (NMR)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 50 mg/mL
ethanol: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N1(C(=O)C=CC1=O)COC(=O)CC

InChI

1S/C8H9NO4/c1-2-8(12)13-5-9-6(10)3-4-7(9)11/h3-4H,2,5H2,1H3

InChI key

YXEWPGYLMHXLPS-UHFFFAOYSA-N

General description

A cell-permeable maleimide compound selectively inhibits Werner syndrome WRN helicase activity (IC50 = 20 µM) over its ATPase and exonuclease activities (19% and <10% inhibition, respectively, at 50 µM), while exhibiting no effect against the helicase activity of two other human RecQ family DNA helicases (RECQ1 and Werner syndrome BLM), Fanconi anemia group J helicase FANCJ, as well as three other E. coli helicases, RecQ, UvrD, and DnaB. The WRN-dependent antiproliferation activity of MIRA-1/NSC 19630 in HeLa 1.2.11 cultures (0% vs. 95% inhibition with or without siRNA-mediated WRN-depletion, respectively, after 48 h 3 µM treatment) is shown to be a direct result of apoptosis induction due to replication forks blockage (20-fold increase in PCNA foci staining after 72 h 2 µM treatment), enhanced DSB (17-fold increase in γ-H2AX foci staining after 72 h 2 µM treatment), and increased S-phase population (from 24% to 42% after 72 h 2 µM treatment). In some cells, MIRA-1/NSC 19630 appears to also exert its apoptotic effect, at least in part, by restoring DNA-binding and transactivation function of some, but not all, p53 mutants.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Aggarwal, M., et al. 2011. Proc. Natl. Acad. Sci USA108, 1525.
Bykov, V.J.N., et al. 2005. J. Biol. Chem.280, 30384.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service