Skip to Content
Merck
All Photos(2)

Key Documents

146800

Millipore

Tryptic Soy Agar - Ready-to-use Settle Plates

Penase, Tween, Thiosulfate, Histidine, Lecithin, ICR plus with lockable lid, plate volume × diam. 30 mL × 90 mm, sterile; γ-irradiated, pkg of 10 plates Triple packed, suitable for air monitoring

Synonym(s):

CASO Agar, Casein Soybean Digest Agar, TSA, TSA w. Penase LTHThio sedi. ICR+ 30 ml, Tryptic Soy Agar with Lecithin, Tween, Histidin, Thiosulfate and Penicillinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217
NACRES:
NA.74
Product 146800 is not currently sold in your country. Contact Technical Service

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; γ-irradiated

form

plate

feature

ready-to-use

composition

Histidine
Lecithin
Penase
Thiosulfate
Tween

packaging

pkg of 10 plates Triple packed
box of 120 ea 1468000120
box of 20 ea 1468000020

technique(s)

active air sampling: suitable
passive air sampling: suitable

plate volume × diam.

30 mL × 90 mm

color

light yellow

pH range

7.1-7.5

application(s)

air monitoring
pharmaceutical

compatibility

for use with MAS-100®

shipped in

ambient

storage temp.

2-25°C

suitability

bacteria
fungi

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Agar + Penase (Penicillinase) + LTHThio sedi. ICRplus in 90 mm settle plates (lockable ) is designed for the determination of the total aerobic microbial count in air via active or passive air monitoring as well as fingerprints of personnel in isolators and clean rooms. It is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The formulation of the Soybean Casein-Digest medium contains combination of casein and soy peptones by supplying organic nitrogen, particularly amino acids and longer-chained peptides. Sodium chloride maintains the osmotic equilibrium and the medium is supplemented with pyruvate to provide an efficient neutralization of hydrogen peroxide residues. Lecithin, polysorbate 80 (Tween®), histidine and sodium thiosulfate are neutralizers to inactivate disinfectants containing active agents such as Alcohol (70% ethanol or isopropyl alcohol), Aldehyde, Chlorides, Dichloroisocyanurate, Glucoprotamine, Peracetic acid, Phenols, quaternary ammonium compounds and Biguanides. The medium is also supplemented with penicillinase (≥ 1000 IU/liter) to neutralize residuals of penicillins, such as benzylpenicillin, ampicillin, amoxycillin, carbenicillin and methicillin. In addition, the plate model (plus or “+”) is supplied with a lockable lid for safe transport after sampling without the risk of losing the lid. For aerobic incubation the plates should be locked in the “CLOSED”-position and for anaerobic or microaerophilic incubation in the “VENT” position.

Application

Tryptic Soy Agar (TSA, Soybean Casein Digest Agar) is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The media finds its application in environmental monitoring by determining the total aerobic microbial count in air through active or passive air monitoring as well as fingerprints of personnel in isolators and clean rooms.

Legal Information

MAS-100 is a registered trademark of MBV AG
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Microbial monitoring; environmental monitoring; interim storage; ICR contact plates; contact plates; TSA w. LTHThio Contact ICR+

Related Content

Settle plates for isolators and critical cleanrooms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service