Skip to Content
Merck
All Photos(2)

Key Documents

1.46050

Millipore

Tryptic Soy Agar - Ready-to-use Settle Plates

Tween®, Lecithin, ICR Settle plate, sterile; irradiated, plate diam. 90 mm, suitable for air monitoring

Synonym(s):

TSA, Soybean Casein Digest Agar

Sign Into View Organizational & Contract Pricing

Select a Size

20 PKG
$91.30
120 PKG
$557.00

$91.30


Estimated to ship on29 May 2025


Request more information

Select a Size

Change View
20 PKG
$91.30
120 PKG
$557.00

About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

$91.30


Estimated to ship on29 May 2025


Request more information

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; irradiated

form

plate (ready-to-use)

feature

ready-to-use
ICR Settle plate

composition

Lecithin
Tween®

packaging

pack of 20-120 plates

plate diam.

90 mm

pH

7.3 ( in H2O)

application(s)

air monitoring
pharmaceutical

compatibility

for use with MAS-100®

storage temp.

2-25°C

suitability

bacteria
fungi

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Agar + LT - ICR 30ml in 90 mm settle plates (non-lockable) is designed for the determination of the total aerobic microbial count in air via active or passive air monitoring as well as fingerprints of personnel in isolators and clean rooms. It is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The formulation of the Soybean Casein-Digest medium contains a combination of casein and soy peptones supplying organic nitrogen, particularly amino acids and longer-chained peptides. Sodium chloride maintains the osmotic equilibrium and the medium is supplemented with pyruvate to provide sufficient inactivation of hydrogen peroxide residues. Lecithin and polysorbate 80 (Tween®) are neutralizers to deactivate residues of disinfectant ingredients like Alcohol (70% ethanol or isopropyl alcohol), Dichloroisocyanurate, Glucoprotamine, Peracetic acid, Biguanides and quaternary ammonium compounds.

Application

Tryptic Soy Agar (TSA, Soybean Casein Digest Agar) is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The medium is used for environmental monitoring by determining the total aerobic microbial count in air through active or passive air monitoring as well as fingerprints of personnel in isolators and clean rooms.

Legal Information

MAS-100 is a registered trademark of MBV AG
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Microbial monitoring; environmental monitoring; interim storage; ICR contact plates; contact plates; TSA w. LTHThio Contact ICR+

Related Content

Detect microbial contamination gases in a few easy and automated steps. The MAS-100 Atmos® compressed gas sampler is simple and safe to use, and compliant with 21 CFR Part 11.

Settle plates for isolators and critical cleanrooms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service