Skip to Content
Merck
All Photos(2)

Key Documents

1.46015

Millipore

Tryptic Soy Agar - Ready-to-use Settle Plates

Tween®, Cysteine, Histidine, Lecithin, Settle plate for long incubation, aseptically filled, plate diam. 90 mm, suitable for air monitoring

Synonym(s):

CASO Agar, Casein Soybean Digest Agar, TSA, Tryptic Soy Agar Settle Plate

Sign Into View Organizational & Contract Pricing

Select a Size

20 PKG
$73.90
120 PKG
$444.00

$73.90


Estimated to ship on30 May 2025


Request more information

Select a Size

Change View
20 PKG
$73.90
120 PKG
$444.00

About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

$73.90


Estimated to ship on30 May 2025


Request more information

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

aseptically filled

form

plate (ready-to-use)

feature

ready-to-use
Settle plate for long incubation

composition

Cysteine
Histidine
Lecithin
Tween®

packaging

pkg of Pack of 20 and 120 Plates

technique(s)

microbiological culture: suitable

plate diam.

90 mm

pH

7.3 ( in H2O)

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with MAS-100®

storage temp.

15-25°C

suitability

bacteria
fungi

General description

Tryptic Soy Agar + LTHC - in 90 mm settle plates with 30 mL filling volume for long incubation is designed for the detection of the total aerobic microbial count of preservative-containing samples or efficacy tests of disinfectants.
TSA medium is used for the cultivation and isolation of a wide range of bacteria, yeasts, and molds. The formulation contains a combination of casein and soy peptones supplying organic nitrogen, particularly amino acids and longer-chained peptides. Sodium chloride maintains the osmotic equilibrium. The medium is also supplemented with high amounts of neutralizers such as lecithin, polysorbate (Tween®) 80, histidine, and Cysteine to inactivate disinfectants or preservatives.

Application

Tryptic Soy Agar + LTHC - in 90 mm settle plates are designed for the determination of the total aerobic microbial count of preservative-containing samples or efficacy tests of disinfectants.

Legal Information

MAS-100 is a registered trademark of MBV AG
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Settle plates for isolators and critical cleanrooms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service