Skip to Content
Merck
All Photos(1)

Key Documents

1.09772

Supelco

COD Test, photometric

photometric, 10-150 mg/L (COD), Spectroquant®

Sign Into View Organizational & Contract Pricing

Select a Size

25 TESTS
$210.00

$210.00


Estimated to ship on20 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
25 TESTS
$210.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$210.00


Estimated to ship on20 May 2025


Request a Bulk OrderRequest more information

Product Name

COD Cell Test (Hg-free), photometric, 10-150 mg/L (COD), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

analyte functional class(es)

chemical oxygen demand (COD)

measuring range

10-150 mg/L (COD)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Oxidation with chromosulfuric acid, determination as chromate)

storage temp.

15-25°C

General description

Method: Photometric, 10.0 - 150.0 mg/L, Spectroquant®

The Chemical Oxygen Demand (COD) is a measure for the amount of oxygen consumed during the oxidation of organic matter, which gives a clear indication for the quality of a water source, it is for sure one of the most important parameters in (waste) water analysis. Our Spectroquant® COD Tests allow you to accurately quantify the of COD levels in your sample.
These special Hg-free COD Test Kits require the samples to be free of Chloride to avoid interference and wrong results. The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.
All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode), which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

We are a global leader in the life science industry and have produced test kits to measure numerous analytes. Water Online spoke with us about advances in measuring chemical oxygen demand.

Learn about the importance of lab water quality on Chemical Oxygen Demand (COD) and Biochemical (or Biological) Oxygen Demand (BOD) testing in water and wastewater analysis. Find in-depth guidance to establish the optimal lab water solution for your environmental and process water testing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service