Skip to Content
Merck
All Photos(1)

Documents

1.00687

Supelco

BOD Test, photometric

photometric, 0.5-3000 mg/L (BOD), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

BOD Cell Test, photometric, 0.5-3000 mg/L (BOD), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 50 tests

analyte functional class(es)

biochemical oxygen demand (BOD)

measuring range

0.5-3000 mg/L (BOD)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (mod. Winkler method)

storage temp.

15-25°C

General description

Method: photometric 0.5 - 3000 mg/l Spectroquant®
The BOD (biochemical oxygen demand) is defined as that mass of oxygen that is consumed by microorganisms to oxidatively degrade the organic substances in a sample. Using a cell test allows for an easy and efficient determination of BOD, following a modified Winkler method. All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.


The Spectroquant® Cell Test for BOD comes with three empty 16 mm round cells as well as all the required reagents to perform the analysis according to the procedure described in the accompanying instruction leaflet.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - STOT RE 2

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service