Skip to Content
Merck

Skip To

COD2000

COD Calibration Standard

Synonym(s):

Chemical oxygen demand standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mL

Estimated to ship on22 June 2026fromTruganina

$262.00

About This Item

NACRES:
NA.24
UNSPSC Code:
77101502

$262.00


Estimated to ship on22 June 2026Details


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


CofA

current certificate can be downloaded

Quality Segment

concentration

2000 mg/L in H2O

format

multi-component solution

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

This CRM is intended for the calibration and validation of analytical instruments used to measure chemical oxygen demand (COD).

Features and Benefits

  • Verified and qualified in an ISO/IEC 17025 and ISO 17034 accredited laboratory
  • Traceable to the SI and higher order standards from NIST
  • Assigned value based on a purity determination by mass balance and gravimetrically prepared value
  • Homogeneity assessed in accordance with ISO Guide 35
  • Available in 500 mL high-density polyethylene bottle to assure safe delivery
  • Accompanied with a certificate of analysis (CoA) having information on the expiry date for an effective audit
  • Prepared using deionized water as a solvent

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
COD500BOD1000BOD200
format

multi-component solution

format

matrix material

format

matrix material

format

matrix material

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

CofA

certificate is enclosed in each package

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

concentration

2000 mg/L in H2O

concentration

in H2O

concentration

in H2O

concentration

in H2O


Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters