Skip to Content
Merck
All Photos(2)

Key Documents

1.06657

Supelco

Sodium sulfite

anhydrous for analysis EMSURE® Reag. Ph Eur

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$88.50
1 KG
$136.00
5 KG
$477.00
50 KG
$4,430.00

$88.50


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
$88.50
1 KG
$136.00
5 KG
$477.00
50 KG
$4,430.00

About This Item

Linear Formula:
Na2SO3
CAS Number:
Molecular Weight:
126.04
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-821-4
NACRES:
NA.21
form:
powder

$88.50


Available to ship on31 March 2025Details


Request a Bulk Order

Agency

reag. Ph. Eur.

Quality Level

product line

EMSURE®

Assay

97.0-100.5% (iodometric)

form

powder

potency

2610 mg/kg LD50, oral (Rat)

impurities

≤0.005% Insoluble matter
≤0.03 meq/g Titrable free base

pH

8.8-10 (20 °C, 50 g/L in H2O)

mp

>500 °C (decomposition)

density

2.63 g/cm3 at 20 °C

bulk density

1480 kg/m3

anion traces

chloride (Cl-): ≤0.02%
thiosulfate (S2O32-): ≤0.1%

cation traces

As: ≤0.0001%
Cu: ≤0.0005%
Fe: ≤0.001%
Pb: ≤0.0005%
Se: ≤0.0004%
Zn: ≤0.001%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[O-]S([O-])=O

InChI

1S/2Na.H2O3S/c;;1-4(2)3/h;;(H2,1,2,3)/q2*+1;/p-2

InChI key

GEHJYWRUCIMESM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

  • Structural Properties and Hydrolysability of Paulownia elongate: The Effects of Pretreatment Methods Based on Acetic Acid and Its Combination with Sodium Sulfite or Sodium Sulfite: This study investigates how acetic acid and sodium sulfite pretreatments affect the structural properties and enzymatic hydrolysis of fast-growing Paulownia elongate. The combination significantly improved cellulose content and enzymatic digestibility, demonstrating potential for efficient sugar production (Wang et al., 2022).
  • Effects of oxygen scavengers (Sodium sulfite, sodium bisulfite, sodium dithionite, and sodium metabisulfite) on growth and accumulation of biomass in the green alga Asterarcys quadricellulare: This research explores the impact of various oxygen scavengers on the biomass growth of the green alga Asterarcys quadricellulare. Sodium sulfite was one of the compounds tested for enhancing biomass accumulation (Sudha et al., 2022).
  • A novel photo-assisted activated persulfate strategy for selective oxidation of ammonia nitrogen to dinitrogen using sodium silicate and sodium sulfite as regulators: This paper presents a new strategy for the selective oxidation of ammonia nitrogen to dinitrogen. Sodium sulfite was used as a regulator to enhance the efficiency of the process (Zhang et al., 2024).
  • Effectiveness of sodium sulfite as an electron acceptor for bioenhanced treatment of salt-containing water produced from ASP flooding: This study examines the role of sodium sulfite as an electron acceptor in the bioenhanced treatment of saline water from alkaline-surfactant-polymer flooding. The findings suggest improved treatment efficiency with sodium sulfite addition (Zhang et al., 2021).

Analysis Note

Assay (iodometric): 97.0 - 100.5 %
Identity: passes test
Appearance of solution: passes test
Insoluble matter: ≤ 0.005 %
Free acid: passes test
Titrable free base: ≤ 0.03 meq/g
Chloride (Cl): ≤ 0.02 %
Thiosulfate (S₂O₃): ≤ 0.1 %
Heavy metals (as Pb): ≤ 0.0005 %
As (Arsenic): ≤ 0.0001 %
Cu (Copper): ≤ 0.0005 %
Fe (Iron): ≤ 0.001 %
Pb (Lead): ≤ 0.0005 %
Se (Selenium): ≤ 0.0004 %
Zn (Zinc): ≤ 0.001 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Protocols

Preparation of a standard solution for Sulfite

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service