Key Documents
TEM-G300F1-CU
Sigma-Aldrich
Gilder Finder Grids G300F1
Copper, size 300 mesh, vial of 100
Select a Size
1 PKG
$70.70
$70.70
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 PKG
$70.70
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
$70.70
Check Cart for Availability
Request a Bulk Order
packaging
vial of 100
size
300 mesh
General description
Gilder Finder Grids G300F1. Copper grids, 300 mesh. The block of 625 (25x25) individual cells is divided into smaller blocks bounded by thicker crenellated bars. Each small block of 25 cells has a central alphabet letter from A to Y for identification purposes.
Choose from one of the most recent versions:
Certificates of Analysis (COA)