Key Documents
931152
Sigma-Aldrich
TEM Grids
Holey Carbon Supported Copper Grids, grid size 300 mesh, hole avg. diam. 100 nm, box of 25
Synonym(s):
TEM grids
Select a Size
1 EA
$368.00
$368.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$368.00
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
$368.00
Check Cart for Availability
Request a Bulk Order
Product Name
Holey Carbon Supported Copper Grids, grid size 300 mesh, hole avg. diam. 100 nm, box of 25
material
carbon film (holey)
copper grid
packaging
box of 25
grid size
300 mesh
hole avg. diam.
100 nm
General description
Lacey carbon film supported TEM grids. This carbon coating consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. I want to know whether the graphene is coated by PMMA or not before transfered to the grid.
    1 answer
    1. Yes, graphene generally should be coated by PMMA before transferred to the grid. And PMMA could be removed after transfer.
      Helpful?