Key Documents
TEM-CF313-50
Sigma-Aldrich
C-flat Holey Carbon Grids (CF-1.2/1.3) for TEM
Copper Grids, size 300 mesh, pack of 50
Select a Size
1 PKG
$534.00
$534.00
Estimated to ship on15 April 2025
Request a Bulk Order
Select a Size
Change View
1 PKG
$534.00
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
$534.00
Estimated to ship on15 April 2025
Request a Bulk Order
packaging
pack of 50
size
300 mesh
General description
C-flat Holey Carbon Grids for TEM, Copper Grids, 300 Mesh, 1.2 micron hole, 1.3 micron spacing, 50/pk. C-flat is a holey carbon support film, manufactured without plastics, resists or other soft materials. As a result, the carbon films are flat, uniform
Choose from one of the most recent versions:
Certificates of Analysis (COA)