Key Documents
TEM-Q350AR1.3-2NM
Sigma-Aldrich
QUANTIFOIL R 1.2/1.3 with Ultra-Thin Carbon
Gold Grids, size 300 mesh, pack of 50
Select a Size
1 PKG
$846.00
$846.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 PKG
$846.00
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
$846.00
Check Cart for Availability
Request a Bulk Order
packaging
pack of 50
size
300 mesh
General description
QUANTIFOIL with circular holes is used in cryoelectron tomographic reconstruction. The roundness of the holesis advantageous with respect to the formation of an icelayer of constant thickness. Assessment of the image quality is easier when foil is incl
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. What is the thickness of the coated carbon layer?
    1 answer
    1. The foil thickness of the coated carbon layer is ~12nm thick
      Helpful?