Key Documents
TEM-76042-40
Sigma-Aldrich
Micronporous Silicon Nitride TEM Window Grids for Cryo-EM Suspension
10nm Thick, pack of 10
Select a Size
1 PKG
$501.00
$501.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 PKG
$501.00
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
$501.00
Check Cart for Availability
Request a Bulk Order
packaging
pack of 10
General description
Silicon nitrode microporous TEM window with single square of 500x500 micron size, and 20 nm thick SiN. Plasma cleanable, less than 0.5nm variation in film thickness, tolerate temperatures above 1000 ˚C, provides flat, insulating and hydrophobic surfaces
Choose from one of the most recent versions:
Certificates of Analysis (COA)