Skip to Content
Merck
All Photos(1)

Key Documents

928348

Sigma-Aldrich

BrettPhos ChemBeads

Synonym(s):

2-(Dicyclohexylphosphino)3,6-dimethoxy-2′,4′,6′-triisopropyl-1,1′-biphenyl

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$239.00

$239.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$239.00

About This Item

Empirical Formula (Hill Notation):
C35H53O2P
CAS Number:
Molecular Weight:
536.77
MDL number:
UNSPSC Code:
12352112
NACRES:
NA.82

$239.00


Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

composition

~ 5 wt.% loading of catalyst

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

SMILES string

P(C4CCCCC4)(C3CCCCC3)c1c(ccc(c1c2c(cc(cc2C(C)C)C(C)C)C(C)C)OC)OC

InChI

1S/C35H53O2P/c1-23(2)26-21-29(24(3)4)33(30(22-26)25(5)6)34-31(36-7)19-20-32(37-8)35(34)38(27-15-11-9-12-16-27)28-17-13-10-14-18-28/h19-25,27-28H,9-18H2,1-8H3

InChI key

WDVGNXKCFBOKDF-UHFFFAOYSA-N

Application

BrettPhos is a dialkylbiaryl phosphine ligand developed by the Buchwald group. It promotes cross-coupling reactions more efficiently and exhibits improved reactivity compared to other catalytic systems.


It can be used in:
  • palladium-catalyzed trifluoromethylation of aryl chlorides
  • Buchwald-Hartwig amination
  • synthesis of 4-aryl and alkyl substituted, N6-alkylated pyridazine-3,6-diamines via a Buchwald protocol
ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

Learn more about ChemBeads products

For larger scale uses, product also available in powdered form (718742)

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service