Skip to Content
Merck
All Photos(1)

Key Documents

931055

Sigma-Aldrich

tBuXPhos Pd G3 ChemBeads

Synonym(s):

[(2-Di-tert-butylphosphino-2′,4′,6′-triisopropyl-1,1′-biphenyl)-2-(2′-amino-1,1′-biphenyl)] palladium(II) methanesulfonate, tBuXPhos-Pd-G3

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$239.00

$239.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$239.00

About This Item

Empirical Formula (Hill Notation):
C42H58NO3PPdS
CAS Number:
Molecular Weight:
794.37
MDL number:
UNSPSC Code:
41116105
NACRES:
NA.82

$239.00


Check Cart for Availability

Request a Bulk Order

Assay

~6 wt.% loading of catalyst

Quality Level

form

solid

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

extent of labeling

~6 wt. % loading of catalyst

SMILES string

O=S([O-][Pd+2]1(P(C(C)(C)C)(C(C)(C)C)C2=CC=CC=C2C3=C(C(C)C)C=C(C(C)C)C=C3C(C)C)[C-]4=C(C5=C(N1)C=CC=C5)C=CC=C4)(C)=O

InChI

1S/C29H45P.C12H10N.CH4O3S.Pd/c1-19(2)22-17-24(20(3)4)27(25(18-22)21(5)6)23-15-13-14-16-26(23)30(28(7,8)9)29(10,11)12;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;/h13-21H,1-12H3;1-6,8-9H,13H2;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

SQQOHCLJPLSJGX-UHFFFAOYSA-M

General description

tBuXPhos Pd G3 is a third generation (G3) Buchwald precatalyst. It is air, moisture and thermally-stable and is highly soluble in a wide range of common organic solvents. It has long life in solutions. tBuXPhos Pd G3 is an excellent reagent for Buchwald-Hartwig cross-coupling reaction. Some of its unique features include lower catalyst loadings, shorter reaction time, efficient formation of the active catalytic species and accurate control of ligand: palladium ratio.

For larger scale uses, product also available in powdered form (762229)

Application

tBuXPhos Pd G3 may be used to synthesize 4-cyano-7-azaindole from 4-chloro-7-azaindole by cyanation reaction. It may also be used for the α-arylation reaction of acetate esters at room temperature.

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

Learn more about ChemBeads products

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service