Skip to Content
Merck
All Photos(8)

Key Documents

718742

Sigma-Aldrich

BrettPhos

greener alternative

98%

Synonym(s):

2-(Dicyclohexylphosphino)3,6-dimethoxy-2′,4′,6′-triisopropyl-1,1′-biphenyl

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$46.40
500 MG
$202.00
1 G
$348.00
5 G
$1,220.00
25 G
$4,520.00

$46.40


In StockDetails


Request a Bulk Order

Select a Size

Change View
100 MG
$46.40
500 MG
$202.00
1 G
$348.00
5 G
$1,220.00
25 G
$4,520.00

About This Item

Empirical Formula (Hill Notation):
C35H53O2P
CAS Number:
Molecular Weight:
536.77
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

$46.40


In StockDetails


Request a Bulk Order

Quality Level

Assay

98%

form

solid

reaction suitability

reaction type: Cross Couplings
reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: Fluorinations

greener alternative product score

old score: 8
new score: 1
Find out more about DOZN™ Scoring

greener alternative product characteristics

Waste Prevention
Atom Economy
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

187-195 °C

functional group

phosphine

greener alternative category

SMILES string

COc1c(P(C2CCCCC2)C3CCCCC3)c(c4c(C(C)C)cc(C(C)C)cc4C(C)C)c(OC)cc1

InChI

1S/C35H53O2P/c1-23(2)26-21-29(24(3)4)33(30(22-26)25(5)6)34-31(36-7)19-20-32(37-8)35(34)38(27-15-11-9-12-16-27)28-17-13-10-14-18-28/h19-25,27-28H,9-18H2,1-8H3

InChI key

WDVGNXKCFBOKDF-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Waste Prevention”, “Atom Economy” and “Design for Energy Efficiency”. Click here to view its DOZN scorecard.

Application

BrettPhos is a dialkylbiaryl phosphine ligand developed by the Buchwald group. It promotes cross-coupling reactions more efficiently and exhibits improved reactivity compared to other catalytic systems.
It can be used in:
  • palladium-catalyzed trifluoromethylation of aryl chlorides[1]
  • Buchwald-Hartwig amination[2]
  • synthesis of 4-aryl and alkyl substituted, N6-alkylated pyridazine-3,6-diamines via a Buchwald protocol[3]
For small scale and high throughput uses, product is also available as ChemBeads (928348)

Features and Benefits

  • White crystalline solid
  • Air- and moisture-stable
  • Thermally stable
  • Highly efficient
  • Wide functional group tolerance
  • Excellent selectivity and conversion

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Facile synthesis of 4-aryl and alkyl substituted, N 6-alkylated pyridazine-3, 6-diamines.
Wlochal J & Bailey A.
Tetrahedron Letters, 56(48), 6791-6794 (2015)
The palladium-catalyzed trifluoromethylation of aryl chlorides.
Cho EJ, et al.
Science, 328(5986), 1679-1681 (2010)
Multi-kilo delivery of AMG 925 featuring a Buchwald?Hartwig amination and processing with insoluble synthetic intermediates.
Affouard C, et al.
Organic Process Research & Development, 19(3), 476-485 (2015)
T M Rangarajan et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 20(44), 14218-14225 (2014-09-23)
We report an unprecedented BrettPhos ligand supported Pd-catalyzed CO bond-forming reaction of activated aryl halides with primary fluoroalkyl alcohols. We demonstrate that the Phosphine ligand (BrettPhos) possesses the property of altering the mechanistic pathway of reductive elimination from nucleophile to
David S Surry et al.
Angewandte Chemie (International ed. in English), 47(34), 6338-6361 (2008-07-30)
Palladium-catalyzed amination reactions of aryl halides have undergone rapid development in the last 12 years, largely driven by the implementation of new classes of ligands. Biaryl phosphanes have proven to provide especially active catalysts in this context. This Review discusses

Related Content

The Buchwald group has developed a series of highly active and versatile palladium precatalysts and biarylphosphine ligands used in cross-coupling reactions for the formation of C-C, C–N, C–O, C–F, C–CF3, and C–S bonds. The ligands are electron-rich, and highly tunable to provide catalyst systems with a diverse scope, high stability and reactivity. Furthermore, the new series of precatalysts are air-, moisture and thermally-stable and display good solubility in common organic solvents. The use of precatalysts ensures the efficient generation of the active catalytic species and allows one to accurately adjust the ligand:palladium ratio. The ligands, precatalysts and methodology developed in the Buchwald group are user friendly and have rendered previously difficult cross couplings reactions, much easier to achieve.

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service