901089
Silver nanoparticles ink for spray printing
Synonym(s):
Ag ink, Conductive ink, Smart’Ink S-CS11101 conductive ink
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Ag
UNSPSC Code:
12141740
Recommended Products
description
Resistivity: 2 mΩ/sq, per 25 μm (coated on PET arcophane AO STS 100 μm using normandy coating, thermal curing at 150 °C for 30 min)
form
liquid
color
dark brown to blue
surface tension
27 mN/m±4 mN/m
viscosity
<5 mPa.s(20 °C)
capacity
20±1 % loading (Nanoparticles loading)
storage temp.
2-8°C
General description
Smart′Ink S-CS11101 conductive Ink developed for the printed electronics is particularly well suited for applications requiring high conductivity and high resolution. This ink, based on silver, is perfectly adapted to design conductive traces on flexible substrates. Also, it is shaped for IoT application which required antennas (HF, UHF).
Smart′Ink S-CS11101 conductive ink shows great performance on spray printer.
Smart′Ink S-CS11101 conductive ink shows great performance on spray printer.
Features and Benefits
Product benefits:
- Very high electrical conductivity.
- Low curing temperature for printing on flexible substrates.
- Non CMR ink.
- Unchanged conductivity on bending.
- Good adhesion on various flexible substrates (PET, Kapton®, …).
Preparation Note
Typical processing guideline:
- Smart′Ink S-CS01130 Conductive Ink is warmed to room temperature (about 10 minutes) and left for 5 minutes in an ultrasonic bath in order to get rid of any aggregates.
- Filtration on a 0.45 μm PTFE filter syringe.
Legal Information
Product of Genes′Ink
Kapton is a registered trademark of E. I. du Pont de Nemours and Company
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
86.0 °F
Flash Point(C)
30 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service