Skip to Content
Merck
All Photos(1)

Key Documents

923567

Sigma-Aldrich

Nano Silver inkjet ink

photonically curable, low resistivity

Synonym(s):

Ag NP-based ink, Silver (Ag) Nanoparticle Ink, Silver nano ink, Smart′Ink S-CS01520

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$931.00

$931.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$931.00

About This Item

Empirical Formula (Hill Notation):
Ag
Molecular Weight:
107.87
UNSPSC Code:
12352103
NACRES:
NA.23

$931.00


Check Cart for Availability

Request a Bulk Order

Quality Level

form

liquid

composition

alcohol / glycol mixture (solvent)

concentration

20 wt. % Ag
20 wt. %±1% (Silver content)

resistivity

5-10 μΩ-cm, after curing by photonic curing for less than 100ms, 3.2 - 6.3 Nb bulk silver
5.4 μΩ-cm, after curing in furnace at 200° C for 5 minutes, 3.4 Nb bulk silver
5.8 μΩ-cm, after curing in furnace at 180° C for 5 minutes, 3.6 Nb bulk silver
6-12 μΩ-cm, after curing by NIR for a few seconds, 3.8 - 7.5 Nb bulk silver
6.6 μΩ-cm, after curing in furnace at 150° C for 5 minutes, 4.1 Nb bulk silver

sheet resistance

13.5 m Ω/sq, 25 mum after curing at 200 Celsius for 5 min on glass
16.5 m Ω/sq, 25 mum after curing at 150 Celsius for 5 min on glass

color

brown, Brown to Very Dark Brown and Faint Grey to Very Dark Grey and Black

surface tension

24 mN/m±3 mN/m

viscosity

12 mPa.s±3%(20 °C) (@100/s)

density

1.1 g/cm3

storage temp.

2-8°C

Application

Nanosilver inkjet ink finds use in the development of emerging energy technologies such as printed solar cells, energy storage devices, and printed heaters. The ink enables the deposition of conductive layers and electrodes required for efficient energy generation, storage, and management. It is also recommended for IoT applications including antennas (HF, UHF) and sensors, OPV and OLED grids, electrodes, touchscreen bezel busbars.
This Smart′Ink S-CS01520 conductive silver ink is particularly well suited for applications requiring high conductivity and high resolution. This ink, based on nano silver particles, is perfectly suited to design conductive traces on flexible substrates. It is also recommended for IoT applications including antennas (HF, UHF) and sensors, OPV and OLED grids, electrodes, touchscreen bezel busbars.

This ink has been developed to allow photonical curing to enable high throughput printing.

Compatible Printing Method: inkjet printing
Compatible Print heads: RICOH (MH5420), XAAR (1003 GS6, 128), KONIKA MINOLTA (KM512SHx, KM1024iMHE, KM1024LHB), FUJIFILM (Dimatix DMP series cartridge)
Compatible substrates: PET (Melinex 406, Melinex 339, Melines 520, Arcophane TCA, Arcophane STS, Folex BG-71), PEN (Teonex), Kapton® , Ceramic
Recommended surface treatment for substrates:
  • Temperature stabilization
  • Argon plasma treatment

Features and Benefits

  • Improved conductivity
  • Superior resolution (increased with substrate surface treatment)
  • Good bending resistance, smooth surface
  • Shiny coating, mirror effect
  • Optimized nano-particle content
  • Curing process compatibility: photonic, NIR, low vacuum oven, thermal curing
  • Cleaning solution: C-FS21511
  • Non-toxic (No CMR ink)

Legal Information

Product of Genes′Ink
Kapton is a registered trademark of E. I. du Pont de Nemours and Company

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F

Flash Point(C)

60 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service