Skip to Content
Merck
All Photos(1)

Key Documents

901083

Sigma-Aldrich

Silver nanoparticles ink for inkjet printing

Synonym(s):

Ag ink, Conductive ink, Smart Jet C47NJET01020 conductive ink

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$559.00

$559.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$559.00

About This Item

Linear Formula:
Ag
UNSPSC Code:
12141740
NACRES:
NA.23

$559.00


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

color

dark brown to blue

surface tension

29 mN/m±3 mN/m

viscosity

13 mPa.s±3 mPa.s(20 °C)

capacity

20±1 % loading (Nanoparticles loading)

storage temp.

2-8°C

General description

Smart′Ink S-CS01120 conductive ink was developed for the printed electronics, is particularly well suited for applications requiring high conductivity and high resolution. This ink, based on silver, is perfectly adapted to design conductive traces on flexible substrates. This non-toxic nano silver ink is suited to design high resolution conductive patterns on flexible substrates for applications such as antennas, sensors, OPV and OLED grids, electrodes and touchscreen bezel busbars.


Smart′Ink S-CS01120 conductive ink shows improved jetting performance and is successfully used on different tools such as Drop on Demand (DOD) inkjet printer FUJIFILM Dimatix, or such as digital micro-dispensing inkjet printer Microdrop and Microfab.

Features and Benefits

Product benefits:
  • Very high electrical conductivity.
  • Low curing temperature for printing on flexible substrates.
  • Non CMR ink.
  • Compatible with various curing process including photonic, IR, low vacuum oven and thermal.
  • Unchanged conductivity on bending.
  • Good adhesion (5B-ASTM D3359) on various flexible substrates (PET, PEN and Kapton).Recommended surface treatment: temperature stabilization, argon plasma.

Preparation Note

  • Qualified printheads: RICOH (MH5420), XAAR (1003 GS6, 128), KONIKA MINOLTA (KM512SHx, KM1024iMHE, KM1024LHB), FUJIFILM (Dimatrix DMP series cartridge)
  • Storage stability: 6 months (fridge)
  • Sintering conditions:
Tunnel furnace: 120°C/5min for 20mW.cm; 150°C/5min for 6mW.cm; 180°C/5min for 3.8mW.cm; 200°C/5min for 2.8mW.cm.
NIR: few seconds for 4-10mW.cm.
Photonic curing: <100 ms for 3-10mW.cm.
Typical processing guideline


  • Smart′Ink S-CS01120 conductive ink is warmed to room temperature (about 10 minutes) and left for 5 minutes in an ultrasonic bath in order to get rid of any aggregates.
  • Filtration on a 0.45 μm PTFE filter syringe to avoid any nozzles clogging.

Legal Information

Product of Genes′Ink.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F

Flash Point(C)

40 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–4 of 4 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. About cleaning the nozzles and tubes of the printer. Will a Terpineol mix like 86480 remove the silver from the nozzles and equipment in general? My system is the MicroDrop MD-K-130/140 nozzles

    1 answer
    1. The effectiveness of Terpineol for cleaning purposes has not been investigated. Product 910643, a Cleaning Solution for Ag ink, is recommended. This Cleaning solution is a mixture of Terpineol and other components. Please see the link below to review this item:
      https://www.sigmaaldrich.com/product/aldrich/910643

      Helpful?

  4. What is the diameter of the silver nanoparticles in this ink?

    1 answer
    1. The nanoparticles in this product have a diameter of 10nm.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service