Skip to Content
Merck
All Photos(1)

Key Documents

907669

Sigma-Aldrich

Silver ink

75 wt%, LIFT printable

Synonym(s):

Ag ink, Laser induced forward transfer

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$383.00

$383.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$383.00

About This Item

Linear Formula:
Ag
UNSPSC Code:
12352103
NACRES:
NA.23

$383.00


Check Cart for Availability

Request a Bulk Order

description

Silver content : 75 wt%
Solvent : Diethylene Glycol Mono Butyl Ether (DGBE)
Surface Tension : 28 dyn/cm (Pendant Drop)

form

paste

particle size

50-70 nm (by Lumisizer)
90-120 nm (by Lumisizer)

viscosity

50,000-100,000 cP (Shear Rate: 1 s-1
250cP - 450 cP (Shear Rate: 1000 s-1))

density

2.85-3.3

storage temp.

15-25°C

Application

This Ag ink product is a conductive high viscous ink based on single-crystal silver nanoparticles in an organic solvent, has been designed for LIFT (Laser Induced Forward Transfer) digital printing and laser sintering.

The ink offers:
  • Uniform and reproducible donor layer with low drying speed
  • Stable accurate jetting in different types of laser systems, wide working window of jetting parameters
  • High speed printing (20-50kHz), allowing high throughput
  • Narrow patterning on plastic and glass substrates (line width ∼50 μm, height ∼0.5 μm, spacing ∼50 μm)
  • Laser sintering of LIFT printed pattern, providing good electrical properties


Fit for:
LIFT digital printing
Additive electronic manufacturing
Printed electronics: RFID, FPD, Sensors

Laser induced forward transfer printing (LIFT) is a non-contact, nozzle-free, one step, direct laser writing process.[1][2]
LIFT can operate under atmospheric conditions, is compatible with low laser fluences, allows the printing of organic and inorganic materials as well as biological elements. The printing process can take place from both liquid and solid phases. It further has a high lateral resolution only defined by the laser spot size. It is a method compatible with inorganic semiconductor sensor fabrication, OLED fabrication and bio printing[3].

Other Notes

Resistivity (4PP): After laser sintering* ≤ 8 μΩ·cm (≤ 5 bulk) for LIFT printed line (∼1-2 μm thick) on plastic substrate
*Parameters should be optimized depending on line geometry and substrate

Storage and Stability

Keep container tightly closed. Store at room temperature, do not heat above 60 °C and do not freeze.

Legal Information

Lumisizer is a trademark of LUM GmbH

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Small molecular weight organic semiconductors are promising for flexible transistor applications in next-gen soft electronics.

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Questions

  1. Hello, could you please disclose how the silver nanoparticles were dispersed in the solution? I am trying to make a paste of copper nanoparticles in a similar solution and would like to know a commercial method for stable dispersion. Thank you

    1 answer
    1. The detailed preparation method for the silver nanoparticles in this product is considered proprietary and cannot be disclosed. Generally, commercial methods for creating a stable dispersion include using surfactants, stabilizers, or dispersing agents. Additionally, techniques such as sonication and high-shear mixing can be effective for achieving stable dispersions. These methods can also be applied when preparing a paste of copper nanoparticles in a similar solution.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service