Skip to Content
Merck
All Photos(1)

Key Documents

574988

Sigma-Aldrich

Ammonium nickel(II) sulfate hexahydrate

99.999% trace metals basis

Synonym(s):

Nickel ammonium sulfate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$201.00

$201.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$201.00

About This Item

Linear Formula:
(NH4)2Ni(SO4)2 · 6H2O
CAS Number:
Molecular Weight:
394.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
crystals and lumps
solubility:
H2O: slightly soluble(lit.)

$201.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99.999% trace metals basis

form

crystals and lumps

reaction suitability

reagent type: catalyst
core: nickel

impurities

≤15.0 ppm Trace Metal Analysis

mp

85-89 °C (lit.)

solubility

H2O: slightly soluble(lit.)

application(s)

battery manufacturing

SMILES string

N.N.[Ni++].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].OS(O)(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/2H3N.Ni.2H2O4S.6H2O/c;;;2*1-5(2,3)4;;;;;;/h2*1H3;;2*(H2,1,2,3,4);6*1H2/q;;+2;;;;;;;;/p-2

InChI key

ZZPAXHDZFDUTLY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tania Campos-Ordonez et al.
Cellular and molecular neurobiology, 39(3), 435-449 (2019-02-17)
Cyclohexane (CHX) is an organic solvent commonly used as a drug-of-abuse. This drug increases the oxidative stress and glial reactivity in the hippocampus, which suggests that this brain region is vulnerable to CHX effects. This study aimed to establish the
K M Cullen
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 42(10), 1383-1391 (1994-10-01)
A reliable new method was developed for detecting neurofibrillary tangles, senile plaques, and neuropil threads in human neural tissue. Excellent morphological definition of the pathological structures was achieved with this procedure without staining normal neuronal and glial elements. The technique
R Barbaroux et al.
The Science of the total environment, 423, 111-119 (2012-03-13)
The extraction of nickel (Ni) from ultramafic soils by phytomining can be achieved using Alyssum murale cultures. This study presents a new process for the valorization of Ni accumulated by this plant through the production of a Ni ammonium disulfate
Heavy metal intensification of DAB-based HRP reaction product.
J C Adams
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 29(6), 775-775 (1981-06-01)
Z C Peng et al.
Brain research bulletin, 36(1), 101-105 (1995-01-01)
Immunostaining of Fos, the nuclear protein encoded by the immediate early gene c-fos, is widely used to reveal the functional activation of neurons. The chemical identity of cells that express c-fos can be investigated with double immunohistochemistry. We report the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service