Skip to Content
Merck
All Photos(1)

Key Documents

456705

Sigma-Aldrich

(DHQ)2AQN

95%

Synonym(s):

Hydroquinine anthraquinone-1,4-diyl diether

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$99.00

$99.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
500 MG
$99.00

About This Item

Empirical Formula (Hill Notation):
C54H56N4O6
CAS Number:
Molecular Weight:
857.05
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$99.00


0 left in stock

Request a Bulk Order

Assay

95%

form

solid

optical activity

[α]20/D +495°, c = 1 in chloroform

mp

175-180 °C (lit.)

functional group

ketone

SMILES string

CC[C@@H]1CN2CC[C@H]1C[C@@H]2[C@H](Oc3ccc(O[C@@H]([C@H]4C[C@@H]5CCN4C[C@H]5CC)c6ccnc7ccc(OC)cc67)c8C(=O)c9ccccc9C(=O)c38)c%10ccnc%11ccc(OC)cc%10%11

InChI

1S/C54H56N4O6/c1-5-31-29-57-23-19-33(31)25-45(57)53(39-17-21-55-43-13-11-35(61-3)27-41(39)43)63-47-15-16-48(50-49(47)51(59)37-9-7-8-10-38(37)52(50)60)64-54(46-26-34-20-24-58(46)30-32(34)6-2)40-18-22-56-44-14-12-36(62-4)28-42(40)44/h7-18,21-22,27-28,31-34,45-46,53-54H,5-6,19-20,23-26,29-30H2,1-4H3/t31-,32-,33+,34+,45-,46-,53-,54-/m1/s1

InChI key

ARCFYUDCVYJQRN-KGHNJIHGSA-N

Application

Superior ligand for asymmetric dihydroxylation reactions of most olefins bearing aliphatic substituents or olefins having heteroatoms in the allylic position.[1]

Legal Information

Sold under license from Rhodia Pharma Solutions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eecker, H. Sharpless, K.B.
Angewandte Chemie (International Edition in English), 35, 448-448 (1996)

Related Content

The Sharpless Lab pursues useful new reactivity and general methods for selectively controlling chemical reactions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service