Skip to Content
Merck
All Photos(1)

Key Documents

204862

Sigma-Aldrich

Vanadium(IV) oxide sulfate hydrate

≥99.99% trace metals basis

Synonym(s):

Vanadyl sulfate hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$203.00
50 G
$623.00

$203.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$203.00
50 G
$623.00

About This Item

Linear Formula:
VOSO4 · xH2O
CAS Number:
Molecular Weight:
163.00 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
solid

$203.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99.99% trace metals basis

form

solid

composition

Degree of hydration, 3-5

reaction suitability

reagent type: catalyst
core: vanadium

impurities

≤100.0 ppm Trace Metal Analysis

mp

105 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=[V++].[H]O[H].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.H2O.O.V/c1-5(2,3)4;;;/h(H2,1,2,3,4);1H2;;/q;;;+2/p-2

InChI key

DKCWBFMZNUOFEM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Vanadium(IV) oxide sulfate hydrate can be used as a starting material to synthesize V2O5 nanoparticles and vanadium-based oxidation catalysts for selective oxidation reactions. It can also be used as a precursor to fabricate various coatingmaterials. For example, it is used as an additive in the formation of black plasma electrolytic oxidation (PEO) coating on aluminum alloy.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark J Hopwood et al.
Scientific reports, 7, 43436-43436 (2017-03-08)
Hydrogen peroxide (H
Alessandro Chieregato et al.
ChemSusChem, 8(2), 398-406 (2014-12-10)
Glycerol surplus from biodiesel synthesis still represents a major problem in the biofuel production chain. Meanwhile, those in the acrylic acid market are looking for new processes that are able to offer viable alternatives to propylene-based production. Therefore, acrylic acid

Questions

1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. How do you calculate the degree of hydration in products 233706 and 204862?

    1 answer
    1. The equation used to determine the degree of hydration for products 233706 and 204862 is the following.((Molecular weight of Metal * Number of Metal Ions) - (% of metal in product as decimal * anhydrous weight of product)) / (% of metal in product as decimal * 18)Note: 18 is the molecular weight of water.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service