Skip to Content
Merck
All Photos(1)

Key Documents

380040

Sigma-Aldrich

Palladium(II) nitrate solution

10 wt. % in 10 wt. % nitric acid, 99.999% trace metals basis

Synonym(s):

Palladium dinitrate solution, Palladous nitrate

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$274.00
50 ML
$752.00

$274.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
$274.00
50 ML
$752.00

About This Item

Linear Formula:
Pd(NO3)2
Molecular Weight:
230.43
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
liquid

$274.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99.999% trace metals basis

form

liquid

reaction suitability

reagent type: catalyst
core: palladium

concentration

10 wt. % in 10 wt. % nitric acid

impurities

≤15.0 ppm Trace Metal Analysis

density

1.118 g/mL at 25 °C

SMILES string

[Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/2NO3.Pd/c2*2-1(3)4;/q2*-1;+2

InChI key

GPNDARIEYHPYAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service