Skip to Content
Merck
All Photos(1)

Key Documents

461490

Sigma-Aldrich

(R)-(+)-N,N-Dimethyl-1-(1-naphthyl)ethylamine

96%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$236.00

$236.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
$236.00

About This Item

Linear Formula:
C10H7CH(CH3)N(CH3)2
CAS Number:
Molecular Weight:
199.29
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$236.00


Estimated to ship on18 May 2025


Request a Bulk Order

Quality Level

Assay

96%

optical activity

[α]23/D +43°, c = 2 in methanol

refractive index

n20/D 1.591 (lit.)

bp

218 °C (lit.)

density

1 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

C[C@@H](N(C)C)c1cccc2ccccc12

InChI

1S/C14H17N/c1-11(15(2)3)13-10-6-8-12-7-4-5-9-14(12)13/h4-11H,1-3H3/t11-/m1/s1

InChI key

AXRXYILTIWBHEP-LLVKDONJSA-N

Application

(R)-(+)-N,N-Dimethyl-1-(1-naphthyl)ethylamine can react with palladium acetate in acetic acid to form cyclopalladated naphthyl adduct.[1]
Base for kinetic resolution of alcohols and acyl halides.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optically active cyclopalladated compounds containing P-chiral ligands. Restricted rotation around the Pd? P bond.
Albert J, et al.
Journal of Organometallic Chemistry, 634(1), 83-89 (2001)
Weidert, P.J. et al.
Liebigs Ann. Chem., 533-533 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service