Skip to Content
Merck
All Photos(1)

Key Documents

127035

Sigma-Aldrich

1-Naphthylmethylamine

97%

Synonym(s):

1-(Aminomethyl)naphthalene, 1-Naphthalenemethylamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$133.00
25 G
$445.00

$133.00


Estimated to ship on28 March 2025


Request a Bulk Order

Select a Size

Change View
5 G
$133.00
25 G
$445.00

About This Item

Linear Formula:
C10H7CH2NH2
CAS Number:
Molecular Weight:
157.21
Beilstein:
2206459
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
97%

$133.00


Estimated to ship on28 March 2025


Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.643 (lit.)

bp

290-293 °C (lit.)

density

1.073 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

NCc1cccc2ccccc12

InChI

1S/C11H11N/c12-8-10-6-3-5-9-4-1-2-7-11(9)10/h1-7H,8,12H2

InChI key

NVSYANRBXPURRQ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

1-Naphthylmethylamine is used as a building block in organic synthesis to produce dyes, pigments, and pharmaceuticals.

1-Naphthylmethylamine increases the induced circular dichroism (ICD) magnitude exhibited by Poly[(4-carboxyphenyl)acetylene][1]. It forms carbamate by reacting with monomethoxypoly(ethylene glycol) succinimido carbonate (mPEG-SC)[2].

Application

  • Photoluminescence Modulation: A study demonstrates the modulation of photoluminescence in Ruddlesden-Popper perovskite using phase distribution regulation, revealing potential applications in optoelectronic devices. 1-Naphthylmethylamine plays a critical role in enhancing the material properties for improved device performance (Zhao et al., 2023).
  • Perovskite Light-Emitting Diodes: Research highlights the dimensional tailoring of quantum wells through ultrahigh vacuum annealing to enhance the efficiency of perovskite light-emitting diodes. This process utilizes 1-Naphthylmethylamine to improve the interlayer electronic properties and device stability, contributing significantly to advancements in LED technology (Yu et al., 2020).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susan D Van Arnum et al.
Journal of pharmaceutical and biomedical analysis, 50(2), 138-143 (2009-05-12)
An UV-HPLC method for the determination of the potency of mPEG-SC, 5 kDa (1) has been developed and validated. Validation acceptance criteria that is typical of small molecule method validation was successfully applied for this method. The method relies on
Kazuhide Morino et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 10(19), 4703-4707 (2004-09-17)
Poly[(4-carboxyphenyl)acetylene] (poly-1) exhibits an intense induced circular dichroism (ICD) in the UV-visible region upon complexation with excess (R)-1-(1-naphthyl)ethylamine ((R)-2), owing to the formation of a predominantly single-handed helical conformation of the polymer backbone. In the presence of a small amount

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service