Skip to Content
Merck
All Photos(2)

Key Documents

115541

Sigma-Aldrich

(R)-(+)-α-Methylbenzylamine

98%

Synonym(s):

(R)-(+)-1-Phenylethylamine

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$160.00
25 ML
$207.00
100 ML
$446.00

$160.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
$160.00
25 ML
$207.00
100 ML
$446.00

About This Item

Linear Formula:
C6H5CH(CH3)NH2
CAS Number:
Molecular Weight:
121.18
Beilstein:
2410916
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$160.00


Check Cart for Availability

Request a Bulk Order

vapor pressure

0.5 mmHg ( 20 °C)

Quality Level

Assay

98%

optical activity

[α]23/D +38°, neat

optical purity

ee: 96% (GLC)

refractive index

n20/D 1.526 (lit.)

bp

187-189 °C (lit.)

density

0.952 g/mL at 20 °C (lit.)

functional group

amine
phenyl

storage temp.

2-8°C

SMILES string

C[C@@H](N)c1ccccc1

InChI

1S/C8H11N/c1-7(9)8-5-3-2-4-6-8/h2-7H,9H2,1H3/t7-/m1/s1

InChI key

RQEUFEKYXDPUSK-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(R)-(+)-α-Methylbenzylamine is a chiral amine that may be used as a substrate to synthesize (S)-α-amino phosphonates,[1] R-α-aminonitriles,[2] N-{(S)-[cyclohexan-(S)-2-ol]}-(R)-α-methylbenzyl amine and N-{(R)-[cyclohexan-(R)-2-ol]}-(R)-α-methylbenzyl amine.[3] In the presence of boric acid, it can be used in the resolution of racemic 1,1′-bi-2-naphthol.[4]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A New, Convenient Method of Resolution of Racemic 1, 1 '-Bi-2-naphthol Using Boric Acid and (R)-(+)-a-Methylbenzylamine
Periasamy M, et al
The Journal of Organic Chemistry, 64(20), 7643-7645 (1999)
Lithium perchlorate/diethylether catalyzed aminocyanation of aldehydes
Heydari A, et al
Tetrahedron Letters, 39(19), 3049-3050 (1998)
New enantiomerically pure aminoalcohols from (R)-a-methylbenzylamine and cyclohexene oxide.
Barbaro P, et al.
Tetrahedron Asymmetry, 7(3), 843-850 (1996)
Lithium perchlorate/diethylether catalyzed aminophosphonation of aldehydes.
Heydari A, et al.
Tetrahedron Letters, 39(37), 6729-6732 (1998)
Lili Men et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 51(5), 2185-2197 (2018-12-12)
VCP-interacting membrane selenoprotein (VIMP), an ER resident selenoprotein, is highly expressed in β-cells, however, the role of VIMP in β-cells has not been characterized. In this study, we studied the relationship between VIMP deficiency and β-cell survival in MIN6 insulinoma

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service