Skip to Content
Merck
All Photos(1)

Key Documents

237450

Sigma-Aldrich

(S)-(−)-1-(1-Naphthyl)ethylamine

≥99%

Synonym(s):

(S)-(−)-α-Methyl-1-naphthalenemethylamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$78.90
5 G
$181.00

$78.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$78.90
5 G
$181.00

About This Item

Linear Formula:
C10H7CH(CH3)NH2
CAS Number:
Molecular Weight:
171.24
Beilstein:
2208024
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$78.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99%

form

liquid

optical activity

[α]20/D −59°, c = 5 in methanol

optical purity

ee: 98% (GLC)

refractive index

n20/D 1.623 (lit.)

bp

153 °C/11 mmHg (lit.)

density

1.067 g/mL at 20 °C (lit.)

functional group

amine

SMILES string

C[C@H](N)c1cccc2ccccc12

InChI

1S/C12H13N/c1-9(13)11-8-4-6-10-5-2-3-7-12(10)11/h2-9H,13H2,1H3/t9-/m0/s1

InChI key

RTCUCQWIICFPOD-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Employed in a preparation of a chiral thiourea solvating agent useful for enantiodiscrimination of α-hydroxy and α-amino acids by 1H NMR.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 63, 7673-7673 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service