Skip to Content
Merck
All Photos(2)

Key Documents

378712

Sigma-Aldrich

(1S)-(+)-Menthyl chloroformate

optical purity ee: 97% (GLC)

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$94.00
25 ML
$364.00

$94.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$94.00
25 ML
$364.00

About This Item

Empirical Formula (Hill Notation):
C11H19ClO2
CAS Number:
Molecular Weight:
218.72
Beilstein:
2414685
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

$94.00


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

optical activity

[α]20/D +83°, c = 1 in chloroform

optical purity

ee: 97% (GLC)

refractive index

n20/D 1.458 (lit.)

bp

108-109 °C/11 mmHg (lit.)

density

1.031 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

CC(C)[C@H]1CC[C@H](C)C[C@@H]1OC(Cl)=O

InChI

1S/C11H19ClO2/c1-7(2)9-5-4-8(3)6-10(9)14-11(12)13/h7-10H,4-6H2,1-3H3/t8-,9+,10-/m0/s1

InChI key

KIUPCUCGVCGPPA-AEJSXWLSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chiral auxiliary in the asymmetric synthesis of quaternary carbon centers.[1]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 26, 5433-5433 (1985)
Mohammad Mehdi Koleini et al.
The journal of physical chemistry. B, 124(1), 224-233 (2019-12-10)
This study aims to elucidate the impact of salinity on the interactions governing the adsorption of polar aromatic oil compounds onto calcite. To this end, molecular dynamics simulations were employed to assess adsorption of a model polar organic molecule (deprotonated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service