Skip to Content
Merck
All Photos(1)

Key Documents

302325

Sigma-Aldrich

2-Chloroethanesulfonyl chloride

96%

Synonym(s):

2-Chloro-1-ethanesulfonyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$82.80
25 G
$239.00

$82.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$82.80
25 G
$239.00

About This Item

Linear Formula:
ClCH2CH2SO2Cl
CAS Number:
Molecular Weight:
163.02
Beilstein:
1751202
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$82.80


Check Cart for Availability

Request a Bulk Order

vapor pressure

15 mmHg ( 84 °C)

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.493 (lit.)

bp

84-86 °C/15 mmHg (lit.)

density

1.56 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

ClCCS(Cl)(=O)=O

InChI

1S/C2H4Cl2O2S/c3-1-2-7(4,5)6/h1-2H2

InChI key

VHCSBTPOPKFYIU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Chloroethanesulfonyl chloride was used in one-pot sulfonylation/intramolecular thia-Michael protocol for the synthesis of 1,5,2-dithiazepine 1,1-dioxides.[1] It was also used in synthesis of vinylsulfonamides with a furan, carbocyclic, semicyclic or acyclic 1,3-diene moiety.[2]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Qin Zang et al.
Heterocycles, 86(2), doi:10-doi:10 (2014-01-05)
A novel one-pot sulfonylation/intramolecular thia-Michael protocol is reported for the synthesis of 1,5,2-dithiazepine 1,1-dioxides. Sulfonylation between cysteine ethyl ester/cysteamine and 2-chloroethanesulfonyl chloride, followed by
Victor O Rogachev et al.
Nature protocols, 1(6), 3076-3087 (2007-04-05)
Vinylsulfonamides with a furan, carbocyclic, semicyclic or acyclic 1,3-diene moiety are synthetized via a domino elimination-amidation reaction of 2-chloroethanesulfonyl chloride. Intramolecular Diels-Alder reaction of these vinylsulfonamides with thermal (toluene, 110 degrees C) or high pressure (dichloromethane, 13 kbar) activation provides

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service