Skip to Content
Merck
All Photos(1)

Key Documents

226173

Sigma-Aldrich

(1S)-(−)-Camphanic chloride

98%

Synonym(s):

(−)-Camphanoyl chloride, (1S)-3-Oxo-4,7,7-trimethyl-2-oxabicyclo[2.2.1]heptane-1-carbonyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$63.60
5 G
$193.00

$63.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$63.60
5 G
$193.00

About This Item

Linear Formula:
C10H13O3Cl
CAS Number:
Molecular Weight:
216.66
Beilstein:
3590860
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$63.60


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

optical activity

[α]23/D −18±1°, c = 2 in dioxane

mp

71-73 °C (lit.)

functional group

acyl chloride
ester

storage temp.

2-8°C

SMILES string

CC1(C)[C@@]2(C)CC[C@@]1(OC2=O)C(Cl)=O

InChI

1S/C10H13ClO3/c1-8(2)9(3)4-5-10(8,6(11)12)14-7(9)13/h4-5H2,1-3H3/t9-,10+/m0/s1

InChI key

PAXWODJTHKJQDZ-VHSXEESVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(1S)-(-)-Camphanic chloride is a chiral derivatizing agent[1]. It can be prepared by reacting (-)-(1S,4R)-camphanic acid with thionyl chloride.[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiral separations of ?-blocking drug substances using derivatization with chiral reagents and normal-phase high-performance liquid chromatography.
Olsen L, et al.
Journal of Chromatography A, 636(2), 231-241 (1993)
(-)-(1S, 4R)-Camphanoyl Chloride.
Gerlach H, et al.
Organic Syntheses, 71, 48-48 (1993)
The determination of the absolute configurations of diastereomers of (S)-camphanoyl 3-hydroxy-5-oxohexanoic acid derivatives by X-ray crystallography.
Kwai-Ming Cheung et al.
Angewandte Chemie (International ed. in English), 41(7), 1198-1202 (2002-12-20)
M Ruud-Christensen et al.
Journal of chromatography, 491(2), 355-366 (1989-07-21)
A reversed-phase high-performance liquid chromatographic assay has been developed for determination of (R)-(--)-and (S)-(+)-proxyphylline in human plasma. The procedure is based on liquid-solid extraction of proxyphylline from plasma followed by derivatization of extracted proxyphylline with (--)-camphanoyl chloride. The ratio between

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service