Skip to Content
Merck
All Photos(1)

Key Documents

331899

Sigma-Aldrich

(1R,2S)-(−)-2-Amino-1,2-diphenylethanol

99%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$65.00
5 G
$297.00

$65.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$65.00
5 G
$297.00

About This Item

Linear Formula:
C6H5CH(NH2)CH(C6H5)OH
CAS Number:
Molecular Weight:
213.28
Beilstein:
2806218
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$65.00


Check Cart for Availability

Request a Bulk Order

Assay

99%

form

solid

optical activity

[α]25/D −7.0°, c = 0.6 in ethanol

mp

142-144 °C (lit.)

functional group

amine
hydroxyl
phenyl

SMILES string

N[C@H]([C@H](O)c1ccccc1)c2ccccc2

InChI

1S/C14H15NO/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,13-14,16H,15H2/t13-,14+/m0/s1

InChI key

GEJJWYZZKKKSEV-UONOGXRCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chiral auxiliary used for Pd(II)-assisted chiral tandem alkylation and carbonylative coupling reactions.[1][2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.J. Masters et al.
The Journal of Organic Chemistry, 56, 5666-5666 (1991)
J.J. Masters et al.
The Journal of Organic Chemistry, 58, 4547-4547 (1993)
Michael L Berger et al.
Bioorganic & medicinal chemistry, 17(9), 3456-3462 (2009-04-07)
We resolved 1,2-diphenylethylamine (DPEA) into its (S)- and (R)-enantiomer and used them as precursors for synthesis of (S)- and (R)-1-(1,2-diphenylethyl)piperidine, flexible homeomorphs of the NMDA channel blocker MK-801. We also describe the synthesis of the dicyclohexyl analogues of DPEA. These

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service