Skip to Content
Merck
All Photos(1)

Key Documents

671223

Sigma-Aldrich

(S,S)-(−)-2-Amino-1,2-diphenylethanol

≥99.5% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$193.00

$193.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$193.00

About This Item

Linear Formula:
C6H5CH(NH2)CH(C6H5)OH
CAS Number:
Molecular Weight:
213.28
Beilstein:
2806217
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$193.00


Check Cart for Availability

Request a Bulk Order

Assay

≥99.5% (HPLC)

optical activity

[α]/D -124±3°, c = 0.87 in ethanol

optical purity

enantiomeric excess: ≥99.0% (HPLC)

mp

116-119 °C

functional group

amine
hydroxyl
phenyl

storage temp.

2-8°C

SMILES string

N[C@H]([C@@H](O)c1ccccc1)c2ccccc2

InChI

1S/C14H15NO/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,13-14,16H,15H2/t13-,14-/m0/s1

InChI key

GEJJWYZZKKKSEV-KBPBESRZSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service