Skip to Content
Merck
All Photos(2)

Key Documents

317500

Sigma-Aldrich

(1S,2R)-(+)-Norephedrine

98%

Synonym(s):

(1S,2R)-2-Amino-1-phenyl-1-propanol, erythro-α-(1-aminoethyl)benzyl alcohol, D-(+)-Norephedrine, Phenylpropanolamine

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$212.00
50 G
$477.00

$212.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$212.00
50 G
$477.00

About This Item

Linear Formula:
C6H5CH(OH)CH(CH3)NH2
CAS Number:
Molecular Weight:
151.21
Beilstein:
2802896
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$212.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

optical activity

[α]20/D +40°, c = 7 in 1 M HCl

mp

51-54 °C (lit.)

functional group

amine
hydroxyl
phenyl

storage temp.

2-8°C

SMILES string

C[C@@H](N)[C@@H](O)c1ccccc1

InChI

1S/C9H13NO/c1-7(10)9(11)8-5-3-2-4-6-8/h2-7,9,11H,10H2,1H3/t7-,9-/m1/s1

InChI key

DLNKOYKMWOXYQA-VXNVDRBHSA-N

General description

(1S,2R)-(+)-Norephedrine is an enantiomer of ephedrine mainly used as a chiral auxiliary for asymmetric synthesis.

Application

(1S,2R)-(+)-Norephedrine may be used in the preparation of dendritic ligands, which on combining with Ru(II) complexes form efficient asymmetric transfer hydrogenation catalysts. It may also be used in the preparation of N-((1S,2R)-1-hydroxy-1-phenylpropan-2-yl)furan-2-carboxamide.[1] This chiral amide can efficiently catalyze the enantioselective ethylation of aromatic and heteroaromatic aldehydes to secondary alcohols in the absence of titanium tetraisopropoxide as promotor.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiral amide from (1S, 2R)-(+)-norephedrine alkaloid in the enantioselective addition of diethylzinc to aryl and heteroaryl aldehydes.
Ananthi N, et al.
Tetrahedron Asymmetry, 20(15), 1731-1735 (2009)
Dendritic catalysts for asymmetric transfer hydrogenation based (1S, 2R)-norephedrine derived ligands.
Liu PN, et al.
Tetrahedron Asymmetry, 14(16), 2481-2485 (2003)
Ana Caroline Costa Sa et al.
Scientific reports, 9(1), 8795-8795 (2019-06-21)
Growing evidence suggests that peripheral factors to the brain driving neuro-inflammation could affect Alzheimer's Disease (AD) and Parkinson's Disease (PD) severity. Herpes simplex virus type 1 (HSV1) infection has been associated with AD while other related viruses, including cytomegalovirus (CMV)
Archana Hinduja
Frontiers in neurology, 11, 71-71 (2020-03-03)
Background: Posterior reversible encephalopathy syndrome (PRES) is an acute neurotoxic syndrome that is characterized by a spectrum neurological and radiological feature from various risk factors. Common neurological symptoms includes headache, impairment in level of consciousness, seizures, visual disturbances, and focal
Stefan W Toennes et al.
Journal of pharmaceutical and biomedical analysis, 179, 113008-113008 (2019-12-02)
Each year, synthetic drugs are occurring in high numbers in the illicit drug market. But data on their pharmacology and toxicology are scarcely available. Therefore, a pilot study was performed to evaluate excretion of 4-fluoroamphetamine (4FA) in humans and identify

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service