Skip to Content
Merck
All Photos(1)

Key Documents

186708

Sigma-Aldrich

(S)-(+)-2-Amino-3-methyl-1-butanol

96%

Synonym(s):

L-Valinol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$69.70
5 G
$164.00
25 G
$792.00

$69.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$69.70
5 G
$164.00
25 G
$792.00

About This Item

Linear Formula:
(CH3)2CHCH(NH2)CH2OH
CAS Number:
Molecular Weight:
103.16
Beilstein:
1719137
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.22

$69.70


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

optical activity

[α]25/D +10°, c = 10 in H2O

optical purity

ee: 95% (GLC)

refractive index

n20/D 1.4548 (lit.)

bp

81 °C/8 mmHg (lit.)

mp

30-32 °C (lit.)

density

0.926 g/mL at 25 °C (lit.)

functional group

amine
hydroxyl

storage temp.

2-8°C

SMILES string

CC(C)[C@H](N)CO

InChI

1S/C5H13NO/c1-4(2)5(6)3-7/h4-5,7H,3,6H2,1-2H3/t5-/m1/s1

InChI key

NWYYWIJOWOLJNR-RXMQYKEDSA-N

Application

(S)-(+)-2-Amino-3-methyl-1-butanol can be used to prepare:
  • Imines and oxazolines by reacting with aldehydes and nitriles, respectively.[1][2]
  • Chiral oxazoline derived multidentate ligands containing cyclophosphazene moiety.[3]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 34, 2015-2015 (1993)
Journal of the Chemical Society. Perkin Transactions 1, 192-192 (1989)
Carla Fernandes et al.
Chirality, 29(8), 430-442 (2017-06-14)
Six chiral derivatives of xanthones (CDXs) were covalently bonded to silica, yielding the corresponding xanthonic chiral stationary phases (XCSPs). The new XCSPs were packed into stainless-steel columns with 150 x 4.6 mm i.d. Moreover, the greening of the chromatographic analysis
Enantioselective palladium catalysed allylic substitution with thienyl oxazoline ligands
Frost CG and Williams JMJ
Tetrahedron Letters, 34(12), 2015-2018 (1993)
Asymmetric synthesis of alpha-substituted o-methoxybenzyl alcohols via stereoselective additions to kinetically resolved o-anisaldehyde (tricarbonyl) chromium
Davies SG and Goodfellow CL
Journal of the Chemical Society. Perkin Transactions 1, 1, 192-194 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service