NMR Deuterated Solvent Properties Reference Chart
Common Solvent Peak Coupling and Chemical Shift Values
Use the chart below to look up the coupling values — JHD and JCD(JCF) distance between multiplet peaks in hertz (Hz) — and chemical shift delta values — ðH(Mult)b and ðc(Mult)b in parts per million (ppm) — of NMR solvents by name or CAS number. Molecular weight, density (25 °C), melting point (mp), and boiling point (bp) are also given.
NMR Solvents Reference Data
NMR Solvent | CAS. No. | Mol. wt | Density (25 °C) | MP (°C)a | BP (°C)a | ðH (Mult)b | JHD | ðC (Mult)b | JCD (JCF) |
---|---|---|---|---|---|---|---|---|---|
Acetic acid-d4 151785
| 1186-52-3 | 64.08 | 1.119 | 15-16 | 115.5 | 11.53 (1) | 178.4 (br) | ||
2.03 (5) | 2 | 20.0 (7) | 20 | ||||||
Acetone- d6 151793 | 666-52-4 | 64.12 | 0.872 | -93.8 | 55.5 | 206.0 (13) | 0.9 | ||
2.04 (5) | 2.2 | 29.8 (7) | 20 | ||||||
Aceto- nitrile-d3 151807 | 2206-26-0 | 44.07 | 0.844 | -48 | 80.7 | 118.2 (br) | |||
1.93 (5) | 2.5 | 1.3 (7) | 21 | ||||||
Benzene- d6 151815 | 1076-43-3 | 84.15 | 0.950 | 6.8 | 79.1 | 7.15 (br) | 128.0 (3) | 24 | |
Chloro-form-d CDCL3 151823 | 865-49-6 | 120.38 | 1.500 | -64 | 60.9 | 7.24 (1) | 77.0 (3) | 32 | |
Cyclo-hexane- d12 151866 | 1735-17-7 | 96.23 | 0.893 | 4-7 | 80.7 | 1.38 (br) | 26.4 (5) | 19 | |
Deuterium oxide HDO 151882 | 7789-20-0 | 20.03 | 1.107 | 3.8 | 101.4 | 4.84 (DSS) | |||
4.81 (TSP) | |||||||||
Dichloro- methane-d2 DCM 444324 | 1665-00-5 | 86.94 | 1.362 | -97 | 40 | 5.32 (3) | 1 | 53.8 (5) | 27 |
N,N-Dimethyl- formamide-d7 DMF 189979 | 4472-41-7 | 80.14 | 1.030 | -61 | 153 | 8.01 (br) | 162.7 (3) | 30 | |
2.91 (5) | 2 | 35.2 (7) | 21 | ||||||
2.74 (5) | 2 | 30.1 (7) | 21 | ||||||
Dimethyl sulfoxide-d6 DMSO 151874 | 2206-27-1 | 84.17 | 1.190 | 16-19 | 189 | 2.49 (5) | 1.7 | 39.5 (7) | 21 |
1,4-Dioxane- d8 186406 | 17647-74-4 | 96.15 | 1.129 | 10-12 | 99 | 3.53 (m) | 66.5 (5) | 22 | |
Ethanol- d6 186414 | 1516-08-1 | 52.11 | 0.892 | -114 | 78 | 5.19 (1) | |||
3.55 (br) | 56.8 (5) | 22 | |||||||
1.11 (m) | 17.2 (7) | 19 | |||||||
Methanol-d4 MEOD 151947 | 811-98-3 | 36.07 | 0.888 | -98 | 65.4 | 4.89 (1) | 49.0 (7) | 21.4 | |
3.30 (5) | 1.7 | ||||||||
2-Propanol-d8 175897 | 22739-76-0 | 68.14 | 0.890 | -89.5 | 82 | 5.12 (1) | |||
3.89 (br) | 62.9 (3) | 21.5 | |||||||
Pyridine- d5 532975 | 7291-22-7 | 84.13 | 1.050 | -42 | 114.4 | 8.71 (br) | 149.9 (3) | 27.5 | |
7.55 (br) | 135.5 (3) | 24.5 | |||||||
7.19 (br) | 123.5 (3) | 25 | |||||||
Tetra-hydro-furan-d8 THF 184314 | 1693-74-9 | 80.16 | 0.985 | -106 | 65-66 | 3.58 (br) | 67.4 (5) | 22 | |
1.73 (br) | 25.3 (br) | 20.5 | |||||||
Toluene-d8 434388 | 2037-26-5 | 100.19 | 0.943 | -93 | 110 | 137.5 (1) | |||
7.09 (m) | 128.9 (3) | 23 | |||||||
7.00 (br) | 128.0 (3) | 24 | |||||||
6.98 (m) | 125.2 (3) | 24 | |||||||
2.09 (5) | 2.3 | 20.4 (7) | 19 | ||||||
Trifluoro- acetic acid-d TFA 152005 | 599-00-8 | 115.03 | 1.493 | -15.4 | 75 | 11.50 (1) | 164.2 (4) | (44) | |
116.6 (4) | (283) | ||||||||
2,2,2-Trifluoro- ethanol-d3 396532 | 77253-67-9 | 103.06 | 1.415 | -44 | 77-80 | 5.02 (1) | 126.3 (4) | (277) | |
3.88 (4 x 3) | 2 (9) | 61.5 (4 x 5) | 22 (36) |
Network error: Failed to fetch