189979
N,N-Dimethylformamide-d7
≥99.5 atom % D
Synonym(s):
DMF-d7, Heptadeutero-N,N-dimethylformamide
About This Item
Recommended Products
isotopic purity
≥99.5 atom % D
Quality Level
Assay
≥99% (CP)
form
liquid
technique(s)
NMR: suitable
impurities
≤0.05% water
water
refractive index
n20/D 1.428 (lit.)
bp
153 °C (lit.)
density
1.03 g/mL at 25 °C (lit.)
mass shift
M+7
SMILES string
[2H]C(=O)N(C([2H])([2H])[2H])C([2H])([2H])[2H]
InChI
1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3/i1D3,2D3,3D
InChI key
ZMXDDKWLCZADIW-YYWVXINBSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Recommended products
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
135.5 °F - closed cup
Flash Point(C)
57.5 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Use this reference table to find the coupling values and chemical shifts of our NMR (deuterated) solvents. Melting and boiling points, molecular weight, density, and CAS number are also listed.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service