Skip to Content
Merck
All Photos(1)

Key Documents

151866

Sigma-Aldrich

Cyclohexane-d12

≥99.6 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D12
CAS Number:
Molecular Weight:
96.23
Beilstein:
1908842
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

isotopic purity

≥99.6 atom % D

Quality Level

Assay

≥99% (CP)

form

liquid

expl. lim.

0.34-6.3 % (lit.)

technique(s)

NMR: suitable

refractive index

n20/D 1.421 (lit.)

bp

80.7 °C (lit.)

mp

6.5 °C (lit.)

density

0.893 g/mL at 25 °C (lit.)

mass shift

M+12

SMILES string

[2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H]

InChI

1S/C6H12/c1-2-4-6-5-3-1/h1-6H2/i1D2,2D2,3D2,4D2,5D2,6D2

InChI key

XDTMQSROBMDMFD-LBTWDOQPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclohexane-d12, a deuterated cyclohexane, is a standard purity solvent useful for routine NMR (Nuclear Magnetic Resonance) studies. Its infrared (vapor and liquid phase in the range of 376-4000cm-1) and Raman (liquid phase) spectral investigations have been reported.[1] Ring inversion of cyclohexane-d12 has been studied by recording its deuterium NMR spectrum in the temperature range -36 to +115°C.[2] It can be prepared by reacting benzene-d6 and deuterium.[3]

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vibrational spectroscopic studies of the phase transitions in cyclohexane-d12 at high pressure.
Haines J and Gilson DFR.
The Journal of Physical Chemistry, 94(11), 4712-4716 (1990)
Rate of chair-to-chair interconversion of cyclohexane-d12 in its neat plastic crystalline phase.
McGrath KJ and Weiss RJ.
The Journal of Physical Chemistry, 97(11), 2497-2499 (1993)
A vibrational study of cyclohexane and some of its isotopic derivatives-I: Raman and infrared spectra and assignments of cyclohexane and cyclohexane-d12.
Wiberg KB and Shrake A.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 27(7), 1139-1151 (1971)
Dynamic deuterium NMR in liquid crystalline solvents: Ring inversion of cyclohexane-d12.
Poupko R and Luz Z.
J. Chem. Phys. , 75(4), 1675-1681 (1981)
Viscosities of benzene-d6 and cyclohexane-d12.
Dixon JA and Schiessler RW.
The Journal of Physical Chemistry, 58(5), 430-432 (1954)

Articles

Use this reference table to find the coupling values and chemical shifts of our NMR (deuterated) solvents. Melting and boiling points, molecular weight, density, and CAS number are also listed.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service