Skip to Content
Merck
All Photos(1)

Documents

186406

Sigma-Aldrich

1,4-Dioxane-d8

≥99 atom % D

Synonym(s):

p-Dioxane-d8, Octadeuterodioxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4D8O2
CAS Number:
Molecular Weight:
96.15
Beilstein:
1307286
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

≥99 atom % D

Assay

≥99% (CP)

form

liquid

technique(s)

NMR: suitable

refractive index

n20/D 1.4198 (lit.)

bp

99 °C (lit.)

mp

12 °C (lit.)

density

1.129 g/mL at 25 °C (lit.)

mass shift

M+8

SMILES string

[2H]C1([2H])OC([2H])([2H])C([2H])([2H])OC1([2H])[2H]

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2/i1D2,2D2,3D2,4D2

InChI key

RYHBNJHYFVUHQT-SVYQBANQSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

1,4-Dioxane-d8 is a deuterated derivative of 1,4-dioxane. It has an isotopic purity of 99atom%D. It is a standard purity solvent suitable for routine NMR analyses (conducted at ambient temperatures where quality is less critical). It is widely employed as a deuterated standard. It participates as a surrogate standard during the activated carbon SPE (solid-phase extraction) and GC-MS-SIM (gas chromatography-mass spectrometry (GC-MS) in selected ion monitoring (SIM) mode) quantification of 1,4-dioxane in drinking water.

Application

  • Neutron Scattering of Residual Hydrogen in 1,4-Dioxane-d8 Liquid: Understanding Measurements with Molecular Dynamics Simulations: This study utilizes 1,4-Dioxane-d8 to enhance neutron scattering measurements, providing crucial data for researchers in material science and analytical chemistry on solvent behavior and molecular interactions (de Almeida et al., 2016).
  • Biodegradation of 1,4-dioxane and transformation of related cyclic compounds by a newly isolated Mycobacterium sp. PH-06: This research highlights the degradation pathways of 1,4-Dioxane-d8, contributing to environmental sciences by offering insights into bioremediation techniques that mitigate solvent impact (Kim et al., 2009).
  • Degradation of 1,4-dioxane and cyclic ethers by an isolated fungus: Explores the fungal degradation of cyclic ethers, including 1,4-Dioxane-d8, underscoring its significance in environmental cleanup and testing standards for academic and pharmaceutical research on organic pollutant breakdown (Nakamiya et al., 2005).

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

41.0 °F - closed cup

Flash Point(C)

5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Method development for the analysis of 1, 4-dioxane in drinking water using solid-phase extraction and gas chromatography-mass spectrometry
Grimmett PE and Munch JW.
Journal of Chromatographic Science, 47(1), 31-39 (2009)

Articles

Use this reference table to find the coupling values and chemical shifts of our NMR (deuterated) solvents. Melting and boiling points, molecular weight, density, and CAS number are also listed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service