Skip to Content
Merck
All Photos(1)

Key Documents

186414

Sigma-Aldrich

Ethanol-d6

anhydrous, ≥99.5 atom % D

Synonym(s):

Ethyl alcohol-d6, Ethyl-d5 alcohol-d, Hexadeuteroethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CD2OD
CAS Number:
Molecular Weight:
52.11
Beilstein:
1699096
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

isotopic purity

≥99.5 atom % D

Assay

≥99% (CP)

form

liquid

expl. lim.

3.5-15 % (lit.)

technique(s)

NMR: suitable

refractive index

n20/D 1.358 (lit.)

bp

78 °C (lit.)

mp

-130 °C (lit.)

density

0.892 g/mL at 25 °C

mass shift

M+6

SMILES string

[2H]OC([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3/i1D3,2D2,3D

InChI key

LFQSCWFLJHTTHZ-LIDOUZCJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethanol-d6 is a deuterated derivative of ethanol. It has an isotopic purity of 99.5atom%D. It is a standard purity solvent suitable for routine NMR analyses (conducted at ambient temperatures where quality is less critical).

Application

Ethanol-d6 may be used as a solvent for the tributylphosphine (PBu3) induced 5-exo cyclization of an o-alkynoylphenol to form deuterated aurone.[1]

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Efficient Synthesis of Aurone Derivatives by the Tributylphosphine-catalyzed Regioselective Cyclization of o-Alkynoylphenols.
Saito K, et al.
Chemistry Letters (Jpn), 44(2), 141-143 (2015)

Articles

Use this reference table to find the coupling values and chemical shifts of our NMR (deuterated) solvents. Melting and boiling points, molecular weight, density, and CAS number are also listed.

Development and validation of a headspace SPME and GC-MS method to determine alcohol content in twenty different Kombucha tea samples.

Protocols

GC/MS Analysis of Ethanol in Low Alcohol Beer on SUPELCOWAX® 10 after Headspace SPME using a 100 μm PDMS Fiber

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Is this a liquid ?

    1 answer
    1. Yes, this product is a neat or pure liquid with a density of 0.892 g/mL, as listed in the 'PROPERTIES' section.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service