Skip to Content
Merck
All Photos(3)

Key Documents

V900034

Sigma-Aldrich

Ammonium sulfate

Vetec, reagent grade, 99%

Synonym(s):

Mascagnite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
reagent grade
form:
powder or crystals

grade

reagent grade

product line

Vetec

Assay

99%

form

powder or crystals

pH

5.0-6.0 (25 °C, 132 g/L)

mp

>280 °C (dec.) (lit.)

density

1.77 g/mL at 25 °C (lit.)

SMILES string

N.N.OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

BFNBIHQBYMNNAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon Gareth Broadley et al.
Acta crystallographica. Section D, Biological crystallography, 68(Pt 11), 1450-1459 (2012-10-24)
MshB, a zinc-based deacetylase, catalyses a step in the mycothiol biosynthetic pathway that involves the deacetylation of 1-O-(2-acetamido-2-deoxy-α-D-glucopyranosyl)-D-myo-inositol (GlcNAc-Ins), via cleavage of an amide bond, to 1-O-(2-amino-2-deoxy-α-D-glucopyranosyl)-D-myo-inositol (GlcN-Ins) and acetate. In this study, MshB was expressed, purified and crystallized. A
Kazunori Sugiura et al.
Biochemical and biophysical research communications, 457(3), 242-248 (2015-01-17)
Intracellular redox state is a critical factor for fundamental cellular functions, including regulation of the activities of various metabolic enzymes as well as ROS production and elimination. Genetically-encoded fluorescent redox sensors, such as roGFP (Hanson, G. T., et al. (2004))
Yohei Y Yamamoto et al.
Journal of molecular biology, 426(14), 2667-2678 (2014-05-27)
Chaperonins are ubiquitous molecular chaperones with the subunit molecular mass of 60kDa. They exist as double-ring oligomers with central cavities. An ATP-dependent conformational change of the cavity induces the folding of an unfolded protein that is captured in the cavity.
James F Hunter et al.
Environmental science & technology, 48(17), 10227-10234 (2014-08-06)
A large number of organic species emitted into the atmosphere contain cycloalkyl groups. While cyclic species are believed to be important secondary organic aerosol (SOA) precursors, the specific role of cyclic moieties (particularly for species with multiple or fused rings)
Adam Kawałek et al.
Aging, 5(1), 67-83 (2013-02-22)
We studied the role of peroxisomal catalase in chronological aging of the yeastHansenula polymorpha in relation to various growth substrates. Catalase-deficient (cat) cells showed a similar chronological life span (CLS) relative to the wild-type control upon growth on carbon and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service