Skip to Content
Merck
All Photos(1)

Key Documents

1.01211

Millipore

Ammonium sulfate

for biochemistry

Synonym(s):

Mascagnite

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€62.40
5 KG
€225.00
50 KG
€1,530.00

€62.40


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 KG
€62.40
5 KG
€225.00
50 KG
€1,530.00

About This Item

Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14
MDL number:
UNSPSC Code:
12352301
EC Index Number:
231-984-1
NACRES:
NA.21

€62.40


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

<1 Pa ( 25 °C)

Quality Level

form

solid

potency

2840 mg/kg LD50, oral (Rat)

pH

5 (100 g/L in H2O)

mp

>280 °C (dec.) (lit.)

solubility

754-764 g/L

density

1.77 g/mL at 25 °C (lit.)

bulk density

850 kg/m3

storage temp.

2-30°C

SMILES string

N.N.OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

BFNBIHQBYMNNAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

for biochemistry

Application

Ammonium sulfate for biochemistry. CAS No. 7783-20-2, EC Number 231-984-1.

Analysis Note

Assay (alkalimetric)≥ 99.5 %Insoluble matter≤ 0.005 %pH-value (5 %; water)4.5 - 5.5Chloride (Cl)≤ 0.0005 %Nitrate (NO₃)≤ 0.001 %Phosphate (PO₄)≤ 0.0005 %As (Arsenic)≤ 0.0001 %Ca (Calcium)≤ 0.001 %Cd (Cadmium)≤ 0.0001 %Cu (Copper)≤ 0.0001 %Fe (Iron)≤ 0.0002 %Pb (Lead)≤ 0.0001 %Zn (Zinc)≤ 0.0001 %Residue on ignition (as sulfate)≤ 0.01 %

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Experience effective immobilization of protein A on wide-pore epoxy-modified silica monoliths for mAb separation, showcasing the efficiency of Chromolith® WP 300 columns in achieving faster and highly reproducible biomolecule separations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service