Skip to Content
Merck
All Photos(2)

Key Documents

1.01217

Supelco

Ammonium sulfate

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Mascagnite

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€44.70
1 KG
€74.40
5 KG
€261.00
25 KG
€1,490.00

€44.70


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
€44.70
1 KG
€74.40
5 KG
€261.00
25 KG
€1,490.00

About This Item

Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14
MDL number:
UNSPSC Code:
12352301
EC Index Number:
231-984-1
NACRES:
NA.21
grade:
ACS reagent
form:
solid
solubility:
754-764 g/L

€44.70


Estimated to ship on29 May 2025


Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

vapor pressure

<1 Pa ( 25 °C)

product line

EMSURE®

Assay

≥99.5% (alkalimetric)

form

solid

potency

2840 mg/kg LD50, oral (Rat)

impurities

≤0.001% Insoluble matter

ign. residue

≤0.005% (as sulfate)

loss

≤0.1% loss on drying, 105 °C

pH

5 (100 g/L in H2O)

mp

>280 °C (dec.) (lit.)

solubility

754-764 g/L

density

1.77 g/mL at 25 °C (lit.)

bulk density

850 kg/m3

anion traces

chloride (Cl-): ≤0.0003%
nitrate (NO3-): ≤0.001%
phosphate (PO43-): ≤0.0005%

cation traces

Al: ≤0.000075%
As: ≤0.00002%
Ca: ≤0.001%
Cd: ≤0.0001%
Cr: ≤0.000075%
Cu: ≤0.0002%
Fe: ≤0.0002%
Mg: ≤0.0005%
Pb: ≤0.0002%
Zn: ≤0.0001%
heavy metals: ≤0.0005%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

N.N.OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

BFNBIHQBYMNNAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (alkalimetric): ≥ 99.5 %
Insoluble matter: ≤ 0.001 %
pH-value (5 %; water, 25 °C): 5.0 - 6.0
Chloride (Cl): ≤ 0.0003 %
Nitrate (NO₃): ≤ 0.001 %
Phosphate (PO₄): ≤ 0.0005 %
Heavy metals (ACS): ≤ 0.0005 %
Heavy metals (as Pb): ≤ 0.0005 %
Al (Aluminium): ≤ 0.000075 %
As (Arsenic): ≤ 0.00002 %
Ca (Calcium): ≤ 0.001 %
Cd (Cadmium): ≤ 0.0001 %
Cr (Chromium): ≤ 0.000075 %
Cu (Copper): ≤ 0.0002 %
Fe (Iron): ≤ 0.0002 %
Mg (Magnesium): ≤ 0.0005 %
Pb (Lead): ≤ 0.0002 %
Zn (Zinc): ≤ 0.0001 %
Residue on ignition (as sulfate): ≤ 0.005 %
Loss on Drying (105 °C): ≤ 0.1 %
Corresponds to ACS,ISO,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

EMSURE® Grade Inorganic Solvents and Reagents offer exceptional purity and quality control, ensuring reliable, compliant results for regulated analyses.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service